Skip to Content
Merck
All Photos(1)

Key Documents

N3911

Sigma-Aldrich

NBI 27914

≥98% (HPLC)

Synonym(s):

5-Chloro-4-(N-(cyclopropyl)methyl-N-propylamino)-2-methyl-6-(2,4,6-trichlorophenyl)aminopyridine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€130.00

€130.00


Estimated to ship on05 April 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€130.00

About This Item

Empirical Formula (Hill Notation):
C18H20Cl4N4
CAS Number:
Molecular Weight:
434.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€130.00


Estimated to ship on05 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: >20 mg/mL at 60 °C
H2O: insoluble

storage temp.

2-8°C

SMILES string

CCCN(CC1CC1)c2nc(C)nc(Nc3c(Cl)cc(Cl)cc3Cl)c2Cl

InChI

1S/C18H20Cl4N4/c1-3-6-26(9-11-4-5-11)18-15(22)17(23-10(2)24-18)25-16-13(20)7-12(19)8-14(16)21/h7-8,11H,3-6,9H2,1-2H3,(H,23,24,25)

InChI key

KNADXBVKFAUMCR-UHFFFAOYSA-N

Gene Information

human ... CRHR1(1394)

Application

NBI 27914 has been used as acorticotropin-releasing hormone receptor (CRHR1) antagonist:
  • to study its effects on steroidogenic gene expression in GD15 mouse testis [1]
  • to study its effects on amyloid β production [2]
  • to study its effects on neurogenic activity of neural stem cells in the adult hippocampus [3]

Biochem/physiol Actions

NBI 27914 is a CRF1 corticotropin-releasing factor receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A transcriptome-wide screen for mRNAs enriched in fetal Leydig cells: CRHR1 agonism stimulates rat and mouse fetal testis steroidogenesis
McDowell E N, et al.
PLoS ONE, 7(10), e47359-e47359 (2012)
CRH Promotes the Neurogenic Activity of Neural Stem Cells in the Adult Hippocampus
Koutmani Y, et al.
Testing, 29(4), 932-945 (2019)
The stress response neuropeptide CRF increases amyloid-beta production by regulating gamma-secretase activity
Park H J, et al.
The Embo Journal, 34(12), 1674-1686 (2015)
Takatoshi Kasai et al.
Cell reports, 30(1), 18-24 (2020-01-09)
The pituitary is a major hormone center that secretes systemic hormones responding to hypothalamus-derived-releasing hormones. Previously, we reported the independent pituitary induction and hypothalamic differentiation of human embryonic stem cells (ESCs). Here, a functional hypothalamic-pituitary unit is generated using human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service