C0961
Corticotropin Releasing Factor Fragment 6-33 human, rat
≥97% (HPLC)
Select a Size
€348.00
Estimated to ship on29 May 2025
Select a Size
About This Item
€348.00
Estimated to ship on29 May 2025
Recommended Products
Quality Level
Assay
≥97% (HPLC)
form
powder
UniProt accession no.
storage temp.
−20°C
SMILES string
CC[C@H](C)[C@H](N)C(=O)N[C@@H](CO)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](Cc1ccccc1)C(=O)N[C@@H](Cc2c[nH]cn2)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCSC)C(=O)N[C@@H](C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](C)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](C)C(=O)N[C@@H](Cc3c[nH]cn3)C(=O)N[C@@H](CO)C(O)=O
InChI
1S/C141H231N41O43S/c1-23-72(16)109(145)136(221)179-100(61-183)135(220)174-93(52-68(8)9)129(214)176-99(58-108(195)196)133(218)173-95(54-70(12)13)134(219)182-111(77(21)185)138(223)178-96(55-78-29-25-24-26-30-78)130(215)175-98(57-80-60-151-64-155-80)131(216)172-92(51-67(6)7)128(213)171-91(50-66(4)5)126(211)163-82(32-28-47-153-141(148)149)119(204)166-88(38-44-107(193)194)124(209)181-110(71(14)15)137(222)177-94(53-69(10)11)127(212)167-87(37-43-106(191)192)122(207)168-89(45-48-226-22)118(203)158-73(17)112(197)160-81(31-27-46-152-140(146)147)116(201)156-74(18)113(198)162-86(36-42-105(189)190)121(206)165-85(35-41-104(144)188)123(208)170-90(49-65(2)3)125(210)159-75(19)114(199)161-84(34-40-103(143)187)120(205)164-83(33-39-102(142)186)117(202)157-76(20)115(200)169-97(56-79-59-150-63-154-79)132(217)180-101(62-184)139(224)225/h24-26,29-30,59-60,63-77,81-101,109-111,183-185H,23,27-28,31-58,61-62,145H2,1-22H3,(H2,142,186)(H2,143,187)(H2,144,188)(H,150,154)(H,151,155)(H,156,201)(H,157,202)(H,158,203)(H,159,210)(H,160,197)(H,161,199)(H,162,198)(H,163,211)(H,164,205)(H,165,206)(H,166,204)(H,167,212)(H,168,207)(H,169,200)(H,170,208)(H,171,213)(H,172,216)(H,173,218)(H,174,220)(H,175,215)(H,176,214)(H,177,222)(H,178,223)(H,179,221)(H,180,217)(H,181,209)(H,182,219)(H,189,190)(H,191,192)(H,193,194)(H,195,196)(H,224,225)(H4,146,147,152)(H4,148,149,153)/t72-,73-,74-,75-,76-,77+,81-,82-,83-,84-,85-,86-,87-,88-,89-,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,109-,110-,111-/m0/s1
InChI key
XHHPINGDTRCKNN-QZXFXOMNSA-N
Gene Information
human ... CRH(1392)
Amino Acid Sequence
Biochem/physiol Actions
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service