Skip to Content
Merck

Skip To

N0271

Filter Membrane

Nitrocellulose, Thin-film membrane, pore size 0.45 μm, diam. 47 mm

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
41104900
PubChem Substance ID:
NACRES:
NB.24
Material:
TF membrane, nitrocellulose membrane, plain white filter
Detection method:
chemiluminescent, colorimetric, fluorometric, radioactive
Pore size:
0.45 μm


Product Name

Filter membranes, nitrocellulose, TF membrane, pore size 0.45 μm, diam. 47 mm

material

TF membrane, nitrocellulose membrane, plain white filter

manufacturer/tradename

Millipore HATF04700

diam.

47 mm

pore size

0.45 μm

detection method

chemiluminescent, colorimetric, fluorometric, radioactive

General description

Biologically inert nitrocellulose membranes containing a small amount of cellulose acetate for improved handling. Autoclavable to 121°C; use below 75°C. Chemically compatible with dilute acids and bases, hydrocarbons, and non-polar liquids. MF-Millipore membranes have water extractables <5%, including a Triton wetting agent TF-Millipore membranes are Triton-free and have even lower water extractables than MF membranes. See Immobilon-NC in the Electrophoresis Equipment section for nitrocellulose blotting membranes.


pictograms

Flame

signalword

Danger

hcodes

pcodes

Hazard Classifications

Flam. Sol. 1

Storage Class

4.1B - Flammable solid hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Dinesh Chandra et al.
Clinical and experimental immunology, 154(2), 224-234 (2008-09-10)
The failure of Leishmania, an intracellular pathogen, to stimulate a pro-inflammatory response following entry into macrophages has been well reported. This occurs in spite of the fact that ligands for the toll-like receptors (TLR) have been recently shown on the



Global Trade Item Number

SKUGTIN
N0271-100EA04061824364132