This product is supplied in 0.3 ug quantities at a minimum concentration of 10 ng/uL. The number of reactions would depend on the specific application. For example, the link below notes using 10 ng as a spike-in amount for validation purposes:
https://www.future-science.com/doi/full/10.2144/btn-2018-0089?rfr_dat=cr_pub++0pubmed&url_ver=Z39.88-2003&rfr_id=ori%3Arid%3Acrossref.org
MBD0026
Microbial community DNA mix
Suitable for PCR, sequencing and NGS, 10 ng/μL
Select a Size
€286.00
Select a Size
About This Item
€286.00
Recommended Products
Quality Level
form
liquid
concentration
10 ng/μL
technique(s)
DNA extraction: suitable
DNA sequencing: suitable
PCR: suitable
shipped in
dry ice
storage temp.
−20°C
Related Categories
General description
The Microbial community DNA mix is composed of defined collection of genomic DNA standards from 10 different bacterial species, with ranging genome sizes and GC content and varying phyla. This collection can serve as standard for benchmarking the performance along the workflow of microbiomics and as a tool to increase reproducibility and allow comparison of results obtained by different labs.
Read here how to use our standards to ensure data integrity for your microbiome research.
Application
Features and Benefits
- Microbial mix for microbiomics and meta- genomics workflow
- Suitable standard for PCR, sequencing and NGS
- Improve Bioinformatics analyses
- Increases reproducibility
- Compare results lab to lab
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
DNA standards enhance metagenomics research integrity, offering precise species study and mixed community standards.
-
I need to know how many reactions we can perfrom using this kit?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service