Skip to Content
Merck
All Photos(1)

Key Documents

MBD0011

Sigma-Aldrich

Inactivated Enterococcus faecalis

Suitable for DNA extraction, PCR, sequencing, next generation sequencing, >10^8 bacteria/ml

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€249.00

€249.00


Check Cart for Availability


Select a Size

Change View
1 ML
€249.00

About This Item

UNSPSC Code:
41105500
NACRES:
NA.51

€249.00


Check Cart for Availability

Quality Level

form

liquid

concentration

>10^8 bacteria/ml

technique(s)

DNA extraction: suitable
DNA sequencing: suitable
PCR: suitable

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

Standardization of sample analysis is currently needed in microbiome genomics research workflow. Lack of standardization can lead to biases and errors in common processes during sample preparation and analysis such as sample amplification, sequencing and bioinformatics analyses. Inactivated Enterococcus faecalis can serve as standard for benchmarking the performance along the workflow of microbiomics or meta-genomics analyses and as a tool to increase reproducibility and allow comparison of results obtained by different labs.

Enterococcus faecalis is a gram positive, facultative anaerobic, non-motile, coci-shaped bacterium. It is a commensal bacterium of the human intestine and a major opportunistic pathogen in immunocompromised and elderly patients. The pathogenesis of E. faecalis infection relies in part on its capacity to colonize the gut. Following disruption of intestinal homeostasis, E. faecalis can overgrow, cross the intestinal barrier, and enter the lymph and bloodstream.

Isolates of E. faecalis that have ended up in strain collections, mainly from clinical infection sources, appear to have acquired resistance to tetracycline and chloramphenicol in the 1950s and 1960s, followed by gentamicin and erythromycin resistance in the 1970s, then ampicillin and vancomycin resistance in the 1980s and 1990s.

Read here how to use our standards to ensure data integrity for your microbiome research.

Application

Inactivated bacteria are provided at >10^8 bacteria/ml concentration in TE buffer pH 8.0. It is recommended to avoid freeze thaw cycles of this product.
Suitable for Quantitative standard for PCR, Sequencing and NGS

Features and Benefits

  • Individual microbial standard for microbiomics and meta-genomics workflow
  • Suitable standard for PCR, sequencing and NGS
  • Improve Bioinformatics analyses
  • Increases reproducibility
  • Compare results lab to lab

Physical form

Liquid -The inactivated bacteria is provided at >10^8 bacteria/ml in TE buffer pH 8.0

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DNA standards enhance metagenomics research integrity, offering precise species study and mixed community standards.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service