Skip to Content
Merck

Skip To

M3755

Myelin Basic Protein Guinea Pig Fragment 68-82

≥97% (HPLC)

Synonym(s):

des-Gly77, des-His78-Myelin Basic Protein Bovine fragment 68-84

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
€ 807,00

About This Item

Empirical Formula (Hill Notation):
C71H113N23O28
CAS Number:
Molecular Weight:
1736.79
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:

€ 807,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)C(CO)NC(=O)CNC(=O)C(N)Cc1ccc(O)cc1)C(=O)N2CCCC2C(=O)NC(CCC(N)=O)C(=O)NC(CCCCN)C(=O)NC(CO)C(=O)NC(CCC(N)=O)C(=O)NC(CCCNC(N)=N)C(=O)NC(CO)C(=O)NC(CCC(N)=O)C(=O)NC(CC(O)=O)C(=O)NC(CCC(O)=O)C(=O)NC(CC(N)=O)C(O)=O

InChI

1S/C71H113N23O28/c1-33(2)25-44(90-65(116)46(30-95)82-54(103)29-81-57(108)36(73)26-34-10-12-35(98)13-11-34)69(120)94-24-6-9-49(94)68(119)88-41(16-20-52(76)101)61(112)83-37(7-3-4-22-72)58(109)92-47(31-96)66(117)86-39(14-18-50(74)99)60(111)84-38(8-5-23-80-71(78)79)59(110)93-48(32-97)67(118)87-40(15-19-51(75)100)62(113)89-43(28-56(106)107)64(115)85-42(17-21-55(104)105)63(114)91-45(70(121)122)27-53(77)102/h10-13,33,36-49,95-98H,3-9,14-32,72-73H2,1-2H3,(H2,74,99)(H2,75,100)(H2,76,101)(H2,77,102)(H,81,108)(H,82,103)(H,83,112)(H,84,111)(H,85,115)(H,86,117)(H,87,118)(H,88,119)(H,89,113)(H,90,116)(H,91,114)(H,92,109)(H,93,110)(H,104,105)(H,106,107)(H,121,122)(H4,78,79,80)

InChI key

ROVJYNCPHLNVSO-UHFFFAOYSA-N

General description

Myelin Basic Protein (MBP) is the second most abundant protein available in the central nervous system (CNS) myelin. It belongs to the intrinsically disordered or conformationally adaptable protein family.

Application

Myelin Basic Protein Guinea Pig Fragment 68-82 has been used for lymphocyte activation in human blood samples. It has also been used as a component of the incomplete Freund′s adjuvant for the induction of autoimmune encephalomyelitis in rats.

Biochem/physiol Actions

Myelin Basic Protein (MBP) is known as an executive molecule of myelin as it is a structural protein that is required for the formation of central nervous system (CNS) myelin. It participates in the adhesion of the cytosolic surfaces of the multi-layered compact myelin. MBP binds with several polyanionic proteins such as tubulin, actin, clathrin, and Ca2+-calmodulin. It binds to several negatively charged lipids on the cytosolic surface of oligodendrocyte membranes and plays a role in the adhesion of these surfaces in the multilayered myelin sheath.
Myelin basic protein fragment that induces experimental allergic encephalomyelitis.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M2295M1891M0689
assay

≥97% (HPLC)

assay

≥80% (SDS-PAGE)

assay

≥90% (SDS-PAGE)

assay

≥95% (SDS-PAGE)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters