Skip to Content
Merck

Skip To

M3167

4-Methylumbelliferyl α-L-arabinopyranoside

≥99% (HPLC), powder

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C15H16O7
CAS Number:
Molecular Weight:
308.28
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352204
EC Number:
273-991-2
MDL number:


Product Name

4-Methylumbelliferyl α-L-arabinopyranoside, ≥99% (HPLC)

Quality Segment

assay

≥99% (HPLC)

form

powder

solubility

pyridine: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CC1=CC(=O)Oc2cc(OC3OCC(O)C(O)C3O)ccc12

InChI

1S/C15H16O7/c1-7-4-12(17)22-11-5-8(2-3-9(7)11)21-15-14(19)13(18)10(16)6-20-15/h2-5,10,13-16,18-19H,6H2,1H3

InChI key

JWIYLOHVJDJZOQ-UHFFFAOYSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
M3167-1G04061826691670
M3167-100MG04061832924557

Questions

Reviews

No rating value

Active Filters