Skip to Content
Merck

Skip To

M2949

MDL 72527

≥98% (HPLC), spermine oxidase SMO inhibitor, powder

Synonym(s):

CPC-200, MDL-72527DA, N,N′-Bis(2,3-butadienyl)-1,4-butanediamine dihydrochloride, N1,N4-Bis(2,3-butadienyl)-1,4-butanediamine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 114,00
25 mg
Check Cart for Availability
€ 448,00
€ 380,80

About This Item

Empirical Formula (Hill Notation):
C12 H20 N2·2 HCl
CAS Number:
Molecular Weight:
265.22
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder

€ 114,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

MDL 72527, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to brown

solubility

H2O: 15 mg/mL, clear

storage temp.

room temp

SMILES string

Cl.Cl.C=C=CCNCCCCNCC=C=C

InChI

1S/C12H20N2.2ClH/c1-3-5-9-13-11-7-8-12-14-10-6-4-2;;/h5-6,13-14H,1-2,7-12H2;2*1H

InChI key

ITVRWVVFVHINOH-UHFFFAOYSA-N

Biochem/physiol Actions

MDL 72527is an inhibitor of polyamine oxidase (spermine oxidase SMO).
MDL 72527is an inhibitor of polyamine oxidase (spermine oxidase SMO). It blocks the production of H2O2 and increases the cells survival. Polyamine catabolism is one of the major causes of the ROS production of prostate cells in general and androgen-induced enhancement of ROS in androgen-dependent CaP cells in particular. Reports suggest that androgen stimulation of prostate cancer results in accumulation of ROS, which lead to progression of cancer, such progression is control by MDL 72527.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML0460SML1069P7505
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (TLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

400

storage temp.

room temp

storage temp.

room temp

storage temp.

2-8°C

storage temp.

-

solubility

H2O: 15 mg/mL, clear

solubility

H2O: 5 mg/mL (clear solution)

solubility

DMSO: 5 mg/mL, clear (warmed)

solubility

H2O: soluble 100 mg/mL, clear, colorless

color

white to brown

color

white to beige

color

white to beige

color

-


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
M2949-5MG04061832722870
M2949-25MG04061832722863

Questions

Reviews

No rating value

Active Filters