Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
5 mg | Check Cart for Availability | € 628,00 |
About This Item
Empirical Formula (Hill Notation):
C19H34FO2P
Molecular Weight:
344.44
UNSPSC Code:
41106300
EC Number:
201-185-2
NACRES:
NA.77
MDL number:
€ 628,00
Check Cart for Availability
Biochem/physiol Actions
An analog of MAFP which has been widely studied as an inhibitor of phospholipases, FAAH, and as a cannabinoid receptor ligand.
Preparation Note
To change the solvent, simply evaporate the methyl acetate under a gentle stream of nitrogen and immediately add the solvent of choice.
Disclaimer
Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.
1 of 1
This Item | ||
|---|---|---|
| assay ≥98% | assay ≥98% | assay ≥97% (TLC) |
| Quality Level 200 | Quality Level 200 | Quality Level 200 |
| form methyl acetate solution | form methyl acetate solution (10 mg/mL) | form liquid |
| shipped in dry ice | shipped in dry ice | shipped in dry ice |
| storage temp. −70°C | storage temp. −70°C | storage temp. −20°C |
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
target_organs
Central nervous system
supp_hazards
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
3.2 °F - closed cup
flash_point_c
-16 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




