Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 ea | Check Cart for Availability | € 548,00 |
About This Item
€ 548,00
material
flat bottom clear polystyrene wells
sterility
non-sterile
feature
low profile
packaging
case of 100 (internal packs of 10)
manufacturer/tradename
Nunc 438733
external L × W
84 mm × 59 mm , w/ lid
maximum volume
10 μL
size
72 wells
surface area
0.2 cm2 , total surface area (cm2)
working volume
10 μL
color
clear
suitability
suitable for (serological applications), suitable for (serotyping, microcytotoxicity and cell cloning studies)
General description
Application
- Serotyping
- Microcytotoxicity
- Cell cloning studies
Features and Benefits
- Limited adsorption of proteins
- Lids have keyed tabs for proper orientation
- Large raised alphanumeric coordinates
- Unique surface treatment to maximize sample and reagent mixing
- MiniTrays for serological applications have slightly smaller well dimensions
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| material flat bottom clear polystyrene wells | material black polystyrene , flat bottom clear wells | material flat bottom clear polystyrene wells | material black , flat bottom black polystyrene wells |
| manufacturer/tradename Nunc 438733 | manufacturer/tradename Nunc 165305 | manufacturer/tradename Nunc 439225 | manufacturer/tradename Nunc 137101 |
| sterility non-sterile | sterility sterile | sterility non-sterile | sterility sterile |
| feature low profile | feature lid, polystyrene/polymer base | feature lid, low profile | feature lid, pinchbar design |
| working volume 10 μL | working volume 200 μL | working volume 10 μL | working volume 250 μL |
| size 72 wells | size 96 wells | size 60 wells | size 96 wells |
Still not finding the right product?
Explore all of our products under Nunc® MicroWell® MiniTrays
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


