Skip to Content
Merck

Skip To

J3205

JNJ-1661010

≥98% (HPLC)

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C19H19N5OS
Molecular Weight:
365.45
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
solid


Quality Segment

assay

≥98% (HPLC)

form

solid

solubility

DMSO: ≥28 mg/mL

originator

Johnson & Johnson

storage temp.

2-8°C

SMILES string

O=C(Nc1ccccc1)N2CCN(CC2)c3nc(ns3)-c4ccccc4

InChI

1S/C19H19N5OS/c25-18(20-16-9-5-2-6-10-16)23-11-13-24(14-12-23)19-21-17(22-26-19)15-7-3-1-4-8-15/h1-10H,11-14H2,(H,20,25)

InChI key

BHBOSTKQCZEAJM-UHFFFAOYSA-N

Biochem/physiol Actions

JNJ-1661010 is a potent and selective fatty acid amide hydrolase (FAAH) inhibitor with >100-fold preferentially selective for FAAH-1 over FAAH-2. FAAH in an integral membrane enzyme within the amidase-signature family. It catalyzes the hydrolysis of several endogenous biologically active lipids and involves in a variety of physiological and pathological processes, including synaptic regulation, regulation of sleep and feeding, locomotor activity, pain and inflammation.
JNJ-1661010 is a potent and selective fatty acid amide hydrolase (FAAH) inhibitor.

Features and Benefits

This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
J3205-5MG04061832947983

Questions

Reviews

No rating value

Active Filters