Skip to Content
Merck

Skip To

I9529

Interleukin-10/Fc Chimera (non-lytic) from mouse

>98% (SDS-PAGE), recombinant, expressed in CHO cells, buffered aqueous solution

Synonym(s):

IL-10:Fc (non-lytic)

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
51111800
NACRES:
NA.32
MDL number:
Form:
buffered aqueous solution
Assay:
>98% (SDS-PAGE)
Biological source:
mouse
Recombinant:
expressed in CHO cells
Mol wt:
98 kDa


biological source

mouse

Quality Level

recombinant

expressed in CHO cells

assay

>98% (SDS-PAGE)

form

buffered aqueous solution

mol wt

98 kDa

packaging

pkg of 10 μg

impurities

endotoxin, tested

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Il10(16153)

Biochem/physiol Actions

IL-10 (interleukin-10) is an anti-inflammatory cytokine. High levels of IL-10 are present in late syphilis.[1] It is secreted by monocytes and lymphocytes.[2] It suppresses Th1 (T helper) cytokines. IL-10 also controls the differentiation and proliferation of T cells, B cells, natural killer cells, antigen-presenting cells and mast cells.[3]
Possesses the biological function of the IL-10 moiety and a prolonged circulating half-life determined by the Fc domain.

Physical form

Solution in 0.2 μm filtered phosphate buffered saline, pH 7.4.

Other Notes

Dimeric fusion protein consisting of mouse IL-10 fused to mutant mouse IgG2a Fc.


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


C Gérard et al.
The Journal of experimental medicine, 177(2), 547-550 (1993-02-01)
Because of its ability to efficiently inhibit in vitro cytokine production by activated macrophages, we hypothesized that interleukin (IL) 10 might be of particular interest in preventing endotoxin-induced toxicity. We therefore examined the effects of IL-10 administration before lipopolysaccharide (LPS)
Maciej Pastuszczak et al.
Human immunology, 76(7), 469-472 (2015-06-24)
Interleukin 10 (IL-10) is a potent anti-inflammatory cytokine. Increased production of IL-10 has been found in late syphilis, presumably creating favorable conditions for bacteria persistence. Single-nucleotide polymorphisms (SNPs) within the promoter of IL-10 gene have been found to influence IL-10
Lucia Elena Alvarado-Arnez et al.
PloS one, 10(9), e0136282-e0136282 (2015-09-05)
Leprosy is a chronic infectious disease that depends on the interplay of several factors. Single nucleotide polymorphisms (SNPs) in host immune related genes have been consistently suggested as participants in susceptibility towards disease. Interleukin-10 (IL-10) is a crucial immunomodulatory cytokine



Global Trade Item Number

SKUGTIN
I9529-10UG04061838071408

Questions

Reviews

No rating value

Active Filters