Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
120 mL | Check Cart for Availability | € 130,00 |
About This Item
form
solution
Quality Segment
shelf life
Expiry date on the label
IVD
for in vitro diagnostic use
application(s)
hematology
histology
storage temp.
room temp
Application
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| form solution | form solution | form solution | form solution |
| application(s) hematology | application(s) hematology | application(s) hematology | application(s) hematology |
| IVD for in vitro diagnostic use | IVD for in vitro diagnostic use | IVD for in vitro diagnostic use | IVD for in vitro diagnostic use |
| shelf life Expiry date on the label | shelf life Expiry date on the label. | shelf life Expiry date on the label | shelf life Expiry date on the label. |
| Quality Level 500 | Quality Level 500 | Quality Level 500 | Quality Level 500 |
| storage temp. room temp | storage temp. room temp | storage temp. room temp | storage temp. room temp |
Still not finding the right product?
Explore all of our products under Tartrazine Solution
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



