Skip to Content
Merck
All Photos(1)

Key Documents

H5042

Sigma-Aldrich

Cytochrome P450 3A4 human

recombinant, expressed in E. coli

Synonym(s):

Cytochrome P450, purified, soluble, 1,4-cineole 2-exo-monooxygenase, Albendazole sulfoxidase, CYP3A3, CYP3A4, Cytochrome P450 NF-25, Cytochrome P450-PCN1

Sign Into View Organizational & Contract Pricing

Select a Size

1000 PMOL
€618.00

€618.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1000 PMOL
€618.00

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54

€618.00


Check Cart for Availability

Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in E. coli

description

purified, soluble

Assay

3.5—25 mg protein/mL (Bradford)

form

liquid

specific activity

12-200 units/mg protein

mol wt

57318.7 g/mol

concentration

3.5-25 mg protein/mL (protein)
50-150 nmol/mL (P450)
6-14 nmol/mg protein (specific content)

technique(s)

cell based assay: suitable

solubility

water: soluble

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CYP3A4(1576)

Looking for similar products? Visit Product Comparison Guide

General description

Research area: IMMUNO AND CKS

Cytochrome P450 3A4 (CYP3A4) indicates a mammalian enzyme (CYP) family 3, subfamily A, gene 4. There are 57 CYP genes and 33 pseudogenes in humans, organized into 18 families and 42 subfamilies. Among these, the 3A4 isoform (CYP3A4) is the most prevalent and is primarily found in the liver and gastrointestinal tract.[1]

Application

Cytochrome P450 3A4 human has been used in circular dichroism analysis.[2]

Biochem/physiol Actions

Cytochrome P450 3A4 (CYP3A4) is one of the isoforms of cytochrome P450 (P450s).[3] Cytochrome P450s (P450s) plays a vital role in processing endogenous compounds and xenobiotics, including pollutants, environmental compounds, and drug molecules. CYP3A4 oxidizes more than 90% of marketed drugs through various metabolic processes.[3][4]CYP3A4 mRNA expression is upregulated in pediatric Crohn’s disease.[5] Moreover, it provides high tolerance against herbicides such as mefenacet, amiprofos-methyl, pyributicarb, norflurazon, trifluralin, pendimethalin, chlorotoluron, and chloroacetamides.

Unit Definition

One unit will convert 1.0 nanomole of testosterone to 6-beta-testosterone per minute at pH 7.4 at 37 °C.

Other Notes

Supplied as purified soluble proteins, rather than microsomal preparations.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modulation of Carbon Nanotubes' Perturbation to the Metabolic Activity of CYP3A4 in the Liver
Zhang Y, et al.
Advances in Functional Materials, 26(6), 841-850 (2016)
Understanding the mechanism of cytochrome P450 3A4: recent advances and remaining problems
Sevrioukova IF, et al.
Journal of Separation Science, 42(9), 3116-3126 (2013)
Understanding the mechanism of cytochrome P450 3A4: recent advances and remaining problems
Sevrioukova IF, et al.
Journal of Separation Science, 42(9), 3116-3126 (2013)
Elisavet Stavropoulou et al.
Frontiers in immunology, 9, 89-89 (2018-02-16)
Cytochromes are expressed in many different tissues of the human body. They are found mostly in intestinal and hepatic tissues. Cytochromes P450 (CYPs) are enzymes that oxidize substances using iron and are able to metabolize a large variety of xenobiotic
Pharmacokinetic-pharmacodynamic consequences and clinical relevance of cytochrome P450 3A4 inhibition
Dresser, et al.
Clinical Pharmacokinetics, 38(1), 41-57 (2000)

Questions

  1. Hello, Could you please provide more details about this product? Specifically, I would like to know the volume of the aliquot. Also, could you provide a more precise concentration in mg/mL?

    1 answer
    1. This product is a solution containing 1 nanomole or 1000 picomoles of Cytochrome P450 3A4. The concentration of several parameters varies from lot to lot and is reported in the product Certificate of Analysis. As noted in the 'PROPERTIES' section, each vial will have a range of 3.5-25 mg protein/mL, 50-150 nmol/mL P450, and 6-14 nmol/mg protein. For example, a recent lot was found to have a P450 concentration of 13 nmol/mg protein and 5.1 mg protein/mL. The volume of the 1000 picomole package for this lot is 1.5 uL. Please see the link below to review a sample or lot-specific Certificate:
      https://www.sigmaaldrich.com/product/sigma/h5042#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service