Skip to Content
Merck
All Photos(1)

Key Documents

H5292

Sigma-Aldrich

Cytochrome P450 2C19 I331V, human

recombinant, expressed in E. coli

Synonym(s):

Cytochrome P450, purified, soluble, CYP2C19 I331V, CYP2C19 Ile331Val

Sign Into View Organizational & Contract Pricing

Select a Size

1000 PMOL
€614.00

€614.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1000 PMOL
€614.00

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54

€614.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in E. coli

description

purified, soluble

form

liquid

specific activity

12-110 units/mg protein

mol wt

54783.3 g/mol

concentration

3.2-25 mg protein/mL (protein)
50-150 nmol/mL (P450)
6-14 nmol/mg protein (specific content)

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CYP2C19(1557)

Looking for similar products? Visit Product Comparison Guide

General description

Supplied as purified soluble proteins, rather than microsomal preparations.

Unit Definition

One unit will convert 1.0 nanomole of omeprazole (6-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole) to 5-hydroxyomeprazole per minute at pH 7.4 at 37 °C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service