Skip to Content
Merck
All Photos(1)

Key Documents

C5235

Sigma-Aldrich

Cytochrome P450 human

4F3B isozyme microsomes, with P450 Reductase and cytochrome b5, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
€356.00

€356.00


Check Cart for Availability


Select a Size

Change View
1 VIAL
€356.00

About This Item

EC Number:
MDL number:
UNSPSC Code:
12161501
NACRES:
NA.47

€356.00


Check Cart for Availability

biological source

human

Quality Level

recombinant

expressed in baculovirus infected insect cells (BTI-TN-5B1-4)

form

solution

mol wt

45-60 kDa

packaging

vial of 0.5 nmol

UniProt accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CYP4F3(4051)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cytochrome P450 is a heterogeneous family of isozymes whose primary function is to oxidize small molecules, both as a function of intermediary metabolism (e.g., fatty acids) and to detoxify exogenous compounds (drugs or toxins). Some isoforms have narrow substrate specificity, while others are promiscuous. The CYP1A1 isoform catalyzes 7-deethylation of ethoxyresorufin. Cytochrome P450 (CYP) plays an important role in detoxifying xenobiotics, cellular metabolism and homeostasis. One of the main mechanisms of drug-drug interactions is the induction or inhibition of these enzymes. CYP enzymes are transcriptionally activated by a variety of xenobiotics and by endogenous substrates via receptor-dependent pathways. Inhibition of these enzymes is a major factor in metabolism-based drug-drug interactions, and many chemotherapeutic medications can cause drug interactions by either inhibiting or inducing the cytochrome p450 enzyme system.
The CYP4F3B isoform catalyzes the 20-hydroxylation of leukotriene B4.

Unit Definition

One unit will reduce 1 nanomole of cytochrome C per minute at pH 7.4 at 37 deg C.

Physical form

Solution in 100 mM potassium phosphate buffer, pH 7.4.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Christmas et al.
The Journal of biological chemistry, 274(30), 21191-21199 (1999-07-20)
Cytochrome P450 4F3 (CYP4F3) catalyzes the inactivation of leukotriene B(4) by omega-oxidation in human neutrophils. To understand the regulation of CYP4F3 expression, we analyzed the CYP4F3 gene and cloned a novel isoform (CYP4F3B) that is expressed in fetal and adult
Xiangrong Zhang et al.
PloS one, 9(4), e94962-e94962 (2014-04-17)
The present study characterized in vitro metabolites of 20(R)-25-methoxyl-dammarane-3β, 12β, 20-triol (20(R)-25-OCH3-PPD) in mouse, rat, dog, monkey and human liver microsomes. 20(R)-25-OCH3-PPD was incubated with liver microsomes in the presence of NADPH. The reaction mixtures and the metabolites were identified
Mirza Bojić et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(9), 1438-1446 (2014-07-06)
Cilengitide is a stable cyclic pentapeptide containing an Arg-Gly-Asp motif responsible for selective binding to αVβ3 and αVβ5 integrins. The candidate drug showed unexpected inhibition of cytochrome P450 (P450) 3A4 at high concentrations, that is, a 15-mM concentration caused attenuation
Dongju Lin et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(10), 1727-1736 (2014-07-16)
Diosbulbin B (DIOB), a furan-containing diterpenoid lactone, is the most abundant component of Dioscorea bulbifera L. (DB), a traditional Chinese medicine herb. Administration of purified DIOB or DB extracts has been reported to cause liver injury in animals. The mechanisms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service