G8129
Guanosine 2′:3′-cyclic monophosphate sodium salt
~97%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C10H11N5O7PNa
CAS Number:
Molecular Weight:
367.19
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
Recommended Products
Assay
~97%
solubility
H2O: soluble
storage temp.
−20°C
SMILES string
[Na].NC1=NC(=O)c2ncn(C3OC(CO)C4OP(O)(=O)OC34)c2N1
InChI
1S/C10H12N5O7P.Na.H/c11-10-13-7-4(8(17)14-10)12-2-15(7)9-6-5(3(1-16)20-9)21-23(18,19)22-6;;/h2-3,5-6,9,16H,1H2,(H,18,19)(H3,11,13,14,17);;
InChI key
HOTMPXGKAYLTBL-UHFFFAOYSA-N
Caution
Do not confuse with the common guanosine 2′- & 3′- monophosphate (mixed isomers).
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Robert P Ryan et al.
Molecular microbiology, 63(2), 429-442 (2007-01-24)
Cyclic di-GMP is a second messenger with a role in regulation of a range of cellular functions in diverse bacteria including the virulence of pathogens. Cellular levels of cyclic di-GMP are controlled through synthesis, catalysed by the GGDEF protein domain
L Jovine et al.
Structure (London, England : 1993), 8(5), 527-540 (2000-05-10)
Bacterial signal recognition particle (SRP), consisting of 4.5S RNA and Ffh protein, plays an essential role in targeting signal-peptide-containing proteins to the secretory apparatus in the cell membrane. The 4.5S RNA increases the affinity of Ffh for signal peptides and
J M Pérez-Cañadillas et al.
Biochemistry, 37(45), 15865-15876 (1998-12-08)
The electrostatic behavior of titrating groups in alpha-sarcin was investigated using 1H NMR spectroscopy. A total of 209 chemical shift titration curves corresponding to different protons in the molecule were determined over the pH range of 3.0-8.5. Nonlinear least-squares fits
Ryan Morgan et al.
Journal of bacteriology, 188(21), 7335-7343 (2006-10-20)
Multiple environmental cues have been shown to trigger biofilm detachment, the transition from surface-attached, highly organized communities known as biofilms to the motile lifestyle. The goal of this study was to identify a gene product involved in sensing environmental cues
K Hamada et al.
Mutation research, 326(1), 71-82 (1995-01-01)
U5 snRNA can induce both transformation and chromosome aberrations of cells. The polypurine tract, GGAGAGGAA, of the RNA has been suggested to participate in both phenomena. In vitro transcription expected to give this polypurine oligoribonucleotide was associated with cleavage of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service