Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200682

Sigma-Aldrich

Anti-Human IgG (Fc specific) antibody, Mouse monoclonal

clone GG-7, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€488.00

€488.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
€488.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

€488.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

GG-7, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: suitable
immunofluorescence: suitable
indirect ELISA: 0.25-0.5 μg/mL using 5μg/mL human IgG for coating.

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Monoclonal Anti-Human IgG (Fc specific) (mouse IgG1 isotype) is derived from the GG-7 hybridoma, produced by the fusion of mouse myeloma cells and splenocytes from mouse immunized with Purified human IgG myeloma proteins covalently coupled to polyaminostyrene (PAS) microbeads. IgGs are the most common Immunoglobulins isotype in blood, lymph fluid, cerebrospinal fluid, and peritoneal fluid. IgGs include four subclasses (IgG1, IgG2, IgG3, and IgG4), they consist of a variable Fab fragment (which includes the antibody recognition site), and a conserved Fc fragment.

Immunogen

Purified human IgG myeloma proteins covalently coupled to polyaminostyrene (PAS) microbeads

Application

Monoclonal Anti-Human IgG (Fc specific) antibody has been used in:
  • ELISA
  • Imprint Immunofixation (IIF)
  • Immunofluorometric Assay (IFMA)
  • Hemagglutination (HA)
  • Hemagglutination Inhibition (HAI)
  • Particle Counting Immunoassay (PACIA)
  • detection of cytoplasmic IgG

Biochem/physiol Actions

The crystallizable fragment (Fc) of IgG has various important functions such as complement fixation, site for rheumatoid factor (autoantibody directed to Fc) attachment, passage through the placental membrane and protein A binding. IgG deficiencies are often associated with various diseases.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and function of human and murine receptors for IgG
Unkeless J C, et al.
Annual Review of Immunology, 6(1), 251-281 (1988)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service