Skip to Content
Merck

Skip To

F5273

Monoclonal Anti-Rat Kappa & Lambda Light Chains−FITC antibody produced in mouse

clone RT-39/RL-6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Monoclonal Anti-Rat Kappa & Lambda Light Chains

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.46
UNSPSC Code:
12352203


biological source

mouse

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

secondary antibodies

clone

RT-39/RL-6, monoclonal

form

buffered aqueous solution

mol wt

antigen 25 kDa

storage condition

protect from light

technique(s)

dot immunobinding: 1:40, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:40

isotype

IgG1/IgG2a

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Mixture from hybridomas RT-39 and RL-6, secreting monoclonal antibodies against rat κ and λ light chains, respectively.
The product is a mixture derived from hybridomas RT-39 and RL-6, secreting monoclonal antibodies against rat κ and λ light chains, respectively.
The product recognizes epitopes located on the rat κ and λ light chains on the various rat immunoglobulin classes and subclasses. The epitope recognized on the κ light chains is shared by both the 1a and 1b allotypes of this molecule. The antibody detects the κ and λ light chains derived from normal serum immunoglobulins or from myeloma proteins. Weak cross-reaction is observed with guinea pig immunoglobulins but not with human, bovine, cat, chicken, dog, goat, horse, mouse, pig, rabbit or sheep immunoglobulins.

Application

Monoclonal Anti-Rat Kappa & Lambda Light Chains-FITC antibody produced in mouse has been used in:
  • fluorescent dot binding immunoassay (FDIBA)
  • immunocytochemistry
  • immunohistochemistry
  • flow cytometry

Biochem/physiol Actions

FITC Monoclonal Anti-Rat kappa and lambda Light Chains may serve as an essential tool in the localization of light chains on rat immunoglobulins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 8.0, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
R9010F5016F0767
technique(s)

dot immunobinding: 1:40, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:40

technique(s)

capture ELISA: suitable, dot blot: suitable, immunocytochemistry: suitable, indirect ELISA: 1:2,000, western blot: suitable using denatured and reduced normal serum or myeloma protein

technique(s)

dot immunobinding: 1:16, particle immunofluorescence: 1:16

technique(s)

dot immunobinding: 1:64, particle immunofluorescence: 1:32

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

storage condition

protect from light

storage condition

-

storage condition

protect from light

storage condition

protect from light

antibody form

purified immunoglobulin

antibody form

ascites fluid

antibody form

purified from hybridoma cell culture

antibody form

purified from hybridoma cell culture

mol wt

antigen 25 kDa

mol wt

antigen 25 kDa

mol wt

-

mol wt

-

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
F5273-.5ML04061837516887

Questions

Reviews

No rating value

Active Filters