Skip to Content
Merck

Skip To

F2053

Fas Intracellular Domain human

>90% (SDS-PAGE), recombinant, expressed in E. coli (N-terminal histidine tagged), buffered aqueous solution

Synonym(s):

APO-1 interacellular domain, CD95 intracellular domain, Fas IC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 517,00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Form:
buffered aqueous solution
Assay:
>90% (SDS-PAGE)
Recombinant:
expressed in E. coli (N-terminal histidine tagged)
Mol wt:
>18.8 kDa

€ 517,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in E. coli (N-terminal histidine tagged)

Quality Level

assay

>90% (SDS-PAGE)

form

buffered aqueous solution

mol wt

>18.8 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FAS(355)

General description

Fas is a key component of apoptosis, and is a cell surface receptor. This gene is localized to human chromosome 10q24.1, and is composed of nine exons and eight introns.[1]

Biochem/physiol Actions

Fas and its ligand play an essential role in apoptosis, by activating the cell death signaling pathway. Mutations in this gene are associated with evasion of apoptosis by cells, leading to tumorigenesis, and prevention of clearance of these cells by the immune system.[1] A polymorphism in this gene results in the production of soluble Fas (sFas), which is implicated in the chronic autoimmune disorder called primary Sjögren′s syndrome (pSS).[2]
Fas intracellular domain, amino acids 191-335, is a widely expressed membrane receptor, which is responsible for controlling tissue homeostasis and immune responses. Fas receptor is activated by Fas ligand causing its trimerization. The aggregated Fas intracellular death domain recruits, via FADD adapter, pro-caspase-8 and forms a DISC (death-inducing signaling complex). Caspase 8, in turn, activates the downstream caspases, such as caspase 3, culminating in apoptosis.
Widely expressed membrane receptor responsible for controlling tissue homeostasis and immune responses.

Physical form

Solution in 50 mM Tris-HCl, pH 8.0, 2 mM EDTA, 2 mM DTT, 300 mM NaCl, 0.05% NP-40, and 10% sucrose.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SRP3037F8799SRP3036
recombinant

expressed in E. coli (N-terminal histidine tagged)

recombinant

expressed in E. coli

recombinant

expressed in baculovirus infected Sf21 cells

recombinant

expressed in CHO cells

assay

>90% (SDS-PAGE)

assay

≥98% (HPLC), ≥98% (SDS-PAGE)

assay

>97% (SDS-PAGE)

assay

≥95% (HPLC), ≥95% (SDS-PAGE)

mol wt

>18.8 kDa

mol wt

17.6 kDa

mol wt

55 kDa (glycosylated form)

mol wt

17.9 kDa

form

buffered aqueous solution

form

lyophilized

form

lyophilized powder

form

lyophilized

UniProt accession no.

P25445

UniProt accession no.

P25445

UniProt accession no.

P25446

UniProt accession no.

P48023

Gene Information

human ... FAS(355)

Gene Information

human ... FAS(355)

Gene Information

mouse ... Fas(14102)

Gene Information

human ... FASLG(356)


Storage Class

10 - Combustible liquids

wgk

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

9002-93-1

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Cheng Jin et al.
Medical science monitor : international medical journal of experimental and clinical research, 21, 630-637 (2015-02-28)
The CD95 gene plays a key role in regulating cell growth and tumor genesis. To date, several publications have focused on the CD95 rs1800682A/G site polymorphism and various types of tumors in Asians; however, this association is still controversial and
B A Treviño-Talavera et al.
Genetics and molecular research : GMR, 13(3), 4831-4838 (2014-07-26)
Primary Sjögren's syndrome (pSS) is a chronic systemic autoimmune disease characterized by lymphocytic infiltration of exocrine glands. Soluble Fas receptor (sFas) has been suggested as a Fas-mediated apoptosis blocker that could impair clonal deletion in infiltrated autoreactive cells. The FAS



Global Trade Item Number

SKUGTIN
F2053-100UG04061838070968

Questions

Reviews

No rating value

Active Filters