Skip to Content
Merck

Skip To

F0850

Fetal Bovine Serum

Synonym(s):

Fetal Bovine Sera, Fetal Calf Serum

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71
eCl@ss:
32160409
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine fetus

sterility

sterile-filtered

product line

FBS Classic

composition

Bovine IgG, ≤1 mg/mL , Hemoglobin, ≤20 mg/dL

origin

USA origin

technique(s)

cell culture | hybridoma: suitable, cell culture | mammalian: suitable

impurities

≤10 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

General description

Fetal Bovine Serum (FBS) is a complex mixture with low and high molecular weight biomolecules exhibiting optimal growth-enhancing and growth-suppressing functions. FBS contains molecules, such as growth factors, proteins, vitamins, trace elements, hormones etc. These molecules are important for the growth and maintenance of cells. It is a common supplement for cell culture media.citationcitation FBS is derived from the calf blood. It is devoid of fibrin and clotting factors. Heat inactivation of FBS blocks the complement system.citation FBS derived from clotted blood is the most widely used undefined supplement in eukaryotic, especially mammalian, cell culture.citationcitation It is typically used with the classical media that were developed in the 1950s and 1960s. These include many popular media such as Minimum Essential Medium (MEM),citation Dulbecco′s Modified Eagles Media (DME, DMEM); Iscove′s Modified Eagles Medium (IMDM),citation M199citation and Roswell Park Memorial Institute medium, RPMI-1640.citation

Application

  • tested for the presence of bacteria, virus, mycoplasma, endotoxin and for hemoglobin content
  • triple filtered with 0.1 micron membrane under aseptic conditions
FBS derived from clotted blood is the most widely used undefined supplement in eukaryotic, especially mammalian, cell culture. It is typically used with the classical media that were developed in the 1950s and 1960s. These include many popular media such as Minimum Essential Medium (MEM); Dulbecco′s Modified Eagles Media (DME, DMEM); Iscove′s Modified Eagles Medium (IMDM); M199 and Roswell Park Memorial Institute medium, RPMI-1640.
Fetal Bovine Serum has also been used as a component of:
  • perivascular-resident macrophage-like melanocytes (PVM/M) growth mediumcitation
  • synthetic oviduct fluid (SOF)citation
  • Eagle′s minimum essential medium (EMEM)citation
  • neurobasal mediumcitation
  • Hyclone McCoys 5 A mediumcitation
  • Hanks mediumcitation

Biochem/physiol Actions

FBS remains a popular media supplement because it provides a wide array of functions in cell culture. FBS delivers nutrients, growth and attachment factors and protects cells from oxidative damage and apoptosis by mechanisms that are difficult to reproduce in serum-free media (SFM) systems.

Analysis Note

  • Tested for the presence of bacteria, virus, and mycoplasma
  • Triple filtered with 0.1 micron membrane under aseptic conditions

Other Notes

Tested for the presence of bacteria, virus, mycoplasma, endotoxin and for hemoglobin content. Triple filtered with 0.1 micron membrane under aseptic conditions.


Still not finding the right product?

Explore all of our products under Fetal Bovine Serum


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
F0850-500ML04061835357864
F0850-100ML04061835357857
F0850-50ML04061835441181

Questions

Reviews

No rating value

Active Filters