Key Documents
EP6137928009
Reference Filter Set
for Eppendorf BioSpectrometer® Fluorescence
Select a Size
1 EA
€3,670.00
€3,670.00
Check Cart for Availability
Select a Size
Change View
1 EA
€3,670.00
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
€3,670.00
Check Cart for Availability
feature
for Eppendorf BioSpectrometer® Fluorescence
manufacturer/tradename
Eppendorf® 6137928009 (Global)
Related Categories
General description
Contains 8 filters (1 blank filter, 3 filters for UV/Vis wavelength systematic error and precision, 3 filters for UV/Vis photometric verification, 1 filter for fluorometric verification)
All features of the Eppendorf BioSpectrometer UV/Vis Reference Filter Set
Additional filter for verifying fluorimetric precision (random error) and linearity
Inspection protocol preprogrammed in device
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf BioSpectrometer is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)