Key Documents
EP5820772000
Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges
F-35-48-17, to hold, 40 x 15 mL conical tubes, with 48 steel sleeves and adapters
Select a Size
1 EA
€1,130.00
€1,130.00
Check Cart for Availability
Select a Size
Change View
1 EA
€1,130.00
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
€1,130.00
Check Cart for Availability
feature
to hold 40 x 15 mL conical tubes
high-speed fixed angle rotor
with 48 steel sleeves and adapters
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5820772000 (Global)
parameter
20,913 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5804/5804R/5810/5810R
General description
A wide selections of rotors to fit the Eppendorf 5804 and 5810 centrifuges.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)