EP5820755008
Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges
S-4-104, for 5810/5810R, aerosol-tight, without buckets
Synonym(s):
Lid for Eppendorf® FA-45-48-11 Rotor
About This Item
Recommended Products
feature
aerosol-tight
for 5810/5810R
high-capacity swing-bucket rotor
without buckets (without buckets)
without buckets
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5820755008 (Global)
parameter
3,214 × g max. RCF
3,900 rpm
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5804/5804R/5810/5810R
Looking for similar products? Visit Product Comparison Guide
General description
- Eppendorf® high-capacity versatile swing-bucket rotor holds tubes, bottles from 0.2 mL to 750 mL as well as plates.
- Features a QuickLock rotor lid for ergonomic lid locking.
- The aerosol-tight lock facilitates safe centrifugation of hazardous samples.
- Mixed loading with 2 plate buckets and 2 round tube buckets allows for increased versatility.
- Unique aerodynamic round bucket design aids in saving energy up to 36% compared to Rotor A-4-81.
- Not supplied with buckets.
- Autoclavable.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service