Skip to Content
Merck
All Photos(1)

Key Documents

EP0030129776

Eppendorf® twin.tec Trace PCR 96 Well Plates

skirt, working volume 250 μL, PCR clean, blue

Synonym(s):

multiplate, multiwell plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NB.15
material:
blue
size:
96 well
sterility:
non-sterile
feature:
DNase free
PCR clean
skirt
Pricing and availability is not currently available.

material

blue

sterility

non-sterile

feature

DNase free
PCR clean
skirt

packaging

pack of 25 ea

manufacturer/tradename

Eppendorf® 0030129776

size

96 well

working volume

250 μL

suitability

suitable for PCR

Looking for similar products? Visit Product Comparison Guide

General description

  • Eppendorf® twin.tec Trace PCR Plate 96, skirted, 150 µL, PCR clean, colorless, 25 plates
  • Laser-engraved lot number and expiration date on each single plate to improve traceability in your laboratory processes
  • Unique laser-engraved optical guiding grid and OptiTrack® matrix for quick orientation when pipetting manually
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid, torque- and heat-resistant polycarbonate frame: No warping, bending or breaking during heat incubation or centrifugation
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
Eppendorf® twin.tec Trace PCR Plate 96, skirted, 150 µL, PCR clean, crystal blue, 25 plates
  • Laser-engraved lot number and expiration date on each single plate to improve traceability in your laboratory processes
  • Unique laser-engraved optical guiding grid and OptiTrack® matrix for quick orientation when pipetting manually
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid, torque- and heat-resistant polycarbonate frame: No warping, bending or breaking during heat incubation or centrifugation
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)

Application

PCR

Features and Benefits

  • Laser-engraved lot number and expiration date on each single plate to improve traceability in your laboratory processes
  • Unique laser-engraved optical guiding grid and OptiTrack® matrix for quick orientation when pipetting manually
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid, torque- and heat-resistant polycarbonate frame: No warping, bending or breaking during heat incubation or centrifugation
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service