CLS6770
Corning® LSE™ mini microcentrifuges
AC/DC input 110 - 240 V, 50/60 Hz, Includes Adapters for US, EU, UK, AU plugs
Synonym(s):
Corning® LSE™ mini microcentrifuges, fixed angle centrifuge rotor
About This Item
Recommended Products
description
8-place rotor that will accept standard 1.5 mL to 2.0 mL microcentrifuge tubes, and adapters are included for 0.5/0.4 mL and 0.2 mL tubes. Also included is a 4-place rotor for PCR strip tubes.
feature
CE compliant
Includes Adapters for US, EU, UK, AU plugs
packaging
case of 1
pack of 1
manufacturer/tradename
Corning 6770
parameter
2000 × g speed
6000 rpm speed
technique(s)
cryopreservation: suitable
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Includes an 8-place rotor that will accept standard 1.5 mL to 2.0 mL microcentrifuge tubes
- Includes adapters for 0.5/0.4 mL and 0.2 mL tubes
- 4-place rotor for PCR strip tubes included
- Compact, lightweight and easy-to-use
- Close the lid to start, press the lid release button to stop
- 8 x 1.5/2.0 mL rotor and 4-place strip tube rotors included
- Fixed speed of 6000 rpm generates 2000 x g
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service