Skip to Content
Merck
All Photos(1)

Key Documents

EP0030127730

Eppendorf® Serological Pipets, Sterile Grade

red, capacity 25 mL, pack of 200 ea (4 bags × 50 pcs.)

Synonym(s):

Pipettes, Serological Pipets, pipettes, pipetting tips

Sign Into View Organizational & Contract Pricing

Select a Size

200 EA
€124.00

€124.00


Check Cart for Availability


Select a Size

Change View
200 EA
€124.00

About This Item

UNSPSC Code:
41121500
NACRES:
NB.04

€124.00


Check Cart for Availability

material

red

sterility

sterile

feature

DNA free
graduations
plugged

packaging

pack of 200 ea (4 bags × 50 pcs.)

manufacturer/tradename

Eppendorf® 0030127730 (Global)

capacity

25 mL

Looking for similar products? Visit Product Comparison Guide

General description

  • Eppendorf® Serological Pipets are manufactured from ultra-pure virgin polystyrene for optimal clarity.
  • Easy-to-read graduations for accurate volume determination.
  • Pipets are color-coded based on sizes for easy identification.
  • Sterility assurance level of 10-6.
  • Compatible with all mechanical and electronic pipette controllers - including Pipet Helper® or Easypet® 3.
  • Free of detectable pyrogens, DNA, RNase and DNase and certified non-cytotoxic.

Application

  • Transfer of liquids
  • Mixing suspensions
  • Combining reagents and chemical solutions
  • Transferring cells for empirical analysis or expansion
  • Layering reagents for creating higher density gradients

Legal Information

Easypet is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Pipet Helper is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service