Skip to Content
Merck
All Photos(1)

Key Documents

EP0030119380

Eppendorf Tube®

capacity 5.0 mL (5.0), PCR clean, Starter Pack, PCR clean, 400 Tubes, 2 Tube-Racks, 8 adapters for rotors with bore for 15 mL conical tubes

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€167.00

€167.00


Check Cart for Availability


Select a Size

Change View
1 EA
€167.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.11

€167.00


Check Cart for Availability

material

cap (plug seal)
colorless cap
colorless tube
conical bottom
polypropylene cap
polypropylene tube
transparent polypropylene

sterility

sterile

feature

PCR clean
graduations
neck thread Push fit
Starter Pack, PCR clean, 400 Tubes, 2 Tube-Racks, 8 adapters for rotors with bore for 15 mL conical tubes
conical shaped bottom with hinged lid

manufacturer/tradename

Eppendorf® 0030119380

parameter

25,000 × g

technique(s)

PCR: suitable

capacity

5.0 mL (5.0)

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf Tubes are the perfect option for working with medium-sized sample volumes!

  • Maximum safety and stability for centrifugation up to 25,000 × g
  • Conical bottoms
  • Easier processing of samples up to 5.0 mL
  • Higher yields in DNA isolation, prep of mastermixes and buffers, and safe cell and tissue lysis
  • Transparent polypropylene, free of plasticizers, biocides or mold release agents
  • Hinged lid for minimized sample evaporation during storage and incubation in a wide range of temperatures from -86°C to 80°C

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Tube is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service