Skip to Content
Merck

Skip To

EMU190851

MISSION® esiRNA

targeting mouse Ppp3cc

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
20 μg
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
€ 212,00
50 μg
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
€ 382,00

About This Item

NACRES:
NA.51
UNSPSC Code:
41105324

€ 212,00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TAAAAGAAGGTCGGGTGGAAGAGGAGGTGGCCTTAAAGATCATCAATGATGGGGCTGCCATCCTGAAGCAGGAGAAGACCATGATAGAGGTGGAGGCTCCGATCACAGTGTGTGGTGATGTTCATGGACAATTCTTTGACCTGATGAAGTTGTTTGAAGTTGGGGGATCACCTAGTAATACTCGCTACCTCTTCCTGGGTGACTATGTGGACAGAGGCTATTTCAGTATAGAGTGTGTGCTGTATCTATGGAGCTTAAAGATTAACCATCCTAAAACATTGTTTCTGCTTCGAGGAAATCATGAATGCAGGCATCTTACAGAGTACTTCACCTTCAAACAGGAATGTCGGATCAAGTATTCAGAGATGGTGTACGATGCGTGCATGCACACTTTCGACTGTCTTCCTCTTGCTGCCCTCTTAAACCAGCAGTTTCTCTGTGTACATGGAGGAATGTCTCCTGAAATTACTTGTTTAGAGGACATTAGGAAATTAGATAGGTTTTCTGAGCCTCC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EMU021891EMU073261EMU076911
esiRNA cDNA target sequence

TAAAAGAAGGTCGGGTGGAAGAGGAGGTGGCCTTAAAGATCATCAATGATGGGGCTGCCATCCTGAAGCAGGAGAAGACCATGATAGAGGTGGAGGCTCCGATCACAGTGTGTGGTGATGTTCATGGACAATTCTTTGACCTGATGAAGTTGTTTGAAGTTGGGGGATCACCTAGTAATACTCGCTACCTCTTCCTGGGTGACTATGTGGACAGAGGCTATTTCAGTATAGAGTGTGTGCTGTATCTATGGAGCTTAAAGATTAACCATCCTAAAACATTGTTTCTGCTTCGAGGAAATCATGAATGCAGGCATCTTACAGAGTACTTCACCTTCAAACAGGAATGTCGGATCAAGTATTCAGAGATGGTGTACGATGCGTGCATGCACACTTTCGACTGTCTTCCTCTTGCTGCCCTCTTAAACCAGCAGTTTCTCTGTGTACATGGAGGAATGTCTCCTGAAATTACTTGTTTAGAGGACATTAGGAAATTAGATAGGTTTTCTGAGCCTCC

esiRNA cDNA target sequence

TGTGTTCATGGTGGACTTTCACCAGAAATACACACACTGGATGATATTAGGAGATTAGATAGATTTAAAGAGCCACCTGCATTTGGACCAATGTGTGACTTGCTATGGTCTGATCCTTCTGAAGACTTTGGAAATGAAAAATCACAAGAACATTTTAGTCATAATACAGTTCGAGGATGTTCTTATTTTTATAACTATCCAGCAGTGTGTGAATTTTTGCAAAACAATAATTTGTTATCGATTATTAGAGCTCATGAAGCTCAAGATGCAGGCTATAGAATGTACAGAAAAAGTCAAACTACAGGGTTTCCTTCATTAATAACAATTTTTTCGGCACCTAATTACTTAGATGTCTACAATAATAAAGCTGCTGTACTAAAGTATGAAAATAATGTGATGAACATTCGACAGTTTAATTGCTCTCCACATCCTTATTGGTTGCCCA

esiRNA cDNA target sequence

GCGACGGCTAGATTTGTTGTGCTGTGTACATTCCCATGGGGTTGATGGGGTTTTGCCCTAGAGCATCGGATTGAAAATTAAAGTTTTGCTGCTTTCTTCCTTTGCTTTTTAGTTGATGCTGATGAAATTAAAAGGCTAGGAAAGAGATTCAAGAAGCTTGATTTGGACAATTCTGGTTCTTTGAGCGTGGAAGAGTTCATGTCTCTGCCTGAGTTACAGCAGAACCCTTTAGTACAGCGGGTAATAGATATATTCGACACAGACGGCAACGGAGAAGTGGACTTCAAAGAATTCATTGAAGGAGTGTCTCAGTTCAGTGTCAAAGGCGATAAGGAACAGAAGTTGAGGTTTGCTTTTCGTATCTATGACATGGATAAAGATGGCTATATTTCCAATGGAGAACTCTTCCAGGTGTTGAAGATGATGGTGGGCAACAAT

esiRNA cDNA target sequence

TACACGGTGGTTTGTCTCCAGAGATTAACACTCTAGATGACATCAGAAAATTAGACCGATTCAAAGAACCACCTGCTTATGGGCCCATGTGTGACATCCTATGGTCAGACCCCCTGGAGGACTTTGGAAATGAGAAGACTCAGGAACATTTCACTCACAACACAGTCAGAGGCTGTTCGTACTTCTACAGTTACCCAGCTGTGTGTGACTTCCTGCAGCACAATAATTTGTTGTCCATACTCCGCGCCCACGAAGCCCAGGATGCAGGGTACCGCATGTACAGGAAAAGCCAAACAACAGGCTTCCCGTCTCTAATTACAATCTTCTCGGCACCAAATTACTTAGATGTGTACAATAACAAAGCTGCAGTGTTGAAGTACGAGAACAATGTGATGAACATCAGGCAGTTCAACTGCTCCCCGCATCCGTACTGGCTCCCAAATTTCATG

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

Gene Information

mouse ... PPP3CC(19057), Ppp3cc(19057)

Gene Information

mouse ... PPP3CB(19056), Ppp3cb(19056)

Gene Information

mouse ... PPP3R1(19058), Ppp3r1(19058)

Gene Information

mouse ... PPP3CA(19055), Ppp3ca(19055)

Ensembl | mouse accession no.

ENSMUSG00000022092

Ensembl | mouse accession no.

ENSMUSG00000021816

Ensembl | mouse accession no.

ENSMUSG00000033953

Ensembl | mouse accession no.

ENSMUSG00000028161

NCBI accession no.

NM_008915

NCBI accession no.

NM_008914

NCBI accession no.

NM_024459

NCBI accession no.

NM_008913

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
EMU190851-50UG04061828695072
EMU190851-20UG04061828745999

Questions

Reviews

No rating value

Active Filters