Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
TGCTCTCACAACTCCACCACCCCAGCTGCTGGAAATCAAGGGCGGATCTCAAAGGCTGCAGCTATCTTTTTACCAGGAATATCTTCCTATGGCTTCCTTTAGCCTGTCTTCTCCACCCCGTTTATTTTTATCTTAAACAGAGAGACAGGAGAATTACTCTTACACTCTGATAGAGTGTTTTCTACCATCTTCCCAGATGGTCCCTGCGCTCGAAGTCAAAGCACTCTGCCTAGTGGC
GenBank accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
mouse ... MCU(215999), Ccdc109a(215999)
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| Gene Information mouse ... MCU(215999), Ccdc109a(215999) | Gene Information mouse ... ADAP2(216991), Adap2(216991) | Gene Information mouse ... TMEM260(218989), 6720456H20Rik(218989) | Gene Information mouse ... UTP25(215193), AA408296(215193) |
| esiRNA cDNA target sequence TGCTCTCACAACTCCACCACCCCAGCTGCTGGAAATCAAGGGCGGATCTCAAAGGCTGCAGCTATCTTTTTACCAGGAATATCTTCCTATGGCTTCCTTTAGCCTGTCTTCTCCACCCCGTTTATTTTTATCTTAAACAGAGAGACAGGAGAATTACTCTTACACTCTGATAGAGTGTTTTCTACCATCTTCCCAGATGGTCCCTGCGCTCGAAGTCAAAGCACTCTGCCTAGTGGC | esiRNA cDNA target sequence CGACTGCCTGGTCTTAAAGGAACAATGGATTCGAGCTAAGTATGAAAGACAGGAGTTTACGGCTATTGACAAAGCTGTCTCACATCCAGGTAACCGAGAAGGGTTCCTGTGGAAGCGAGGACGGGACAACGCACAGTTCCTGAGAAGGCGGTTTGTCCTTCTGTCGAGGGAAGGCCTCCTGAAGTACTACACCAAGGAGGAGGGTAAAGCCCCAAAGGCTGTCATCAGCATCAAGGACTTAAACGCCACCTTCCAGACAGAGAAGATAGGACATCCCCACGGGCTGCAGATCACCTACAGGAAAGAGGGCCACACCAGGAACCTGTTTGTTTATCATGACAGTGGGAAGGAGATAGTGGATTGGTTCAATGCTCTCCGTGCAGCCCGCCTGCAATACCTGAAATTGGCCTTTCCT | esiRNA cDNA target sequence ATTACTGCGAGGGCTTGAGACCAGATGTTTCACTGGTGGATCAAGAAATGATGACCTATGAGTGGTACTTACCCAAGATGGCGAGGCATTTGCCAGGAGTGCACTTTCCTGGGGACCGGTGGAACCCTGTGGAAGGAGTGTTACCCAGTGGGATGGTCACATTCAACCTCTATCACTTTCTTGAAATGAATAAGCAAAAAGAAACATTTGTTTGCATAGGAATTCATGAAGGCGACCCAACCTGGAAAAAGGACTATTCACTTTGGCCATGGGGATCTTGTGACAAATTGGTGCCTTCAAAGATTGTATTCAACCCAGAGGAGTGGATTGAACGCACAAGAGCCATCTATAACTGGACTGAGGCCTATGAAAGATTTGGTCCATCTTCTTGGGAATCGGTGGCCAATGAGGAGATGTGGCAAGCAAGAATGAAG | esiRNA cDNA target sequence CTTCCTGTCTTCCGTTGAGCTGCTCATCATTGACCAGGCTGACATTTACCTGATGCAGAACTGGGAACATGTCTTGCATTTGATGAACCACATGAACCTGCTGCCCTTGGACTCACACGGGGTGGACTTCTCGAGAGTCCGAATGTGGAGCCTCAATAACTGGTCCAAGTACTACCGACAGACACTGCTCTTTGGTGCCCTGCAGGATGCCCAGATCAACTCTGTGTTCAACAAGCACTGCATCAATGCTCAAGGCCAGGTGGCTGTGAGGAATGTCCCCATGACGGGCTCCATCAGTCATGTGTTGGTGCAGCTCCCTCACGTCTTCCAGAGAATGGAAGCACAAGACCTATCTTCAGTGATCGATGCCAGGTTTCACTTTTTCATAAACAAGATTTTGCCTCAGTACCGGGATGCTGTCATGTCTCACACACTCATCTATGTCCCCTCCTACTTTGACTTTGTGCGTCTCCGAAAT |
| product line MISSION® | product line MISSION® | product line MISSION® | product line MISSION® |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
| NCBI accession no. | NCBI accession no. | NCBI accession no. | NCBI accession no. |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
Storage Class
12 - Non Combustible Liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| EMNC004331-20UG | 04061828816002 |
| EMNC004331-50UG | 04061828795499 |
