Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
description
Powered by Eupheria Biotech
Quality Level
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
GGCTGGCAATGATGAAAACTACTCGGCTGAGCTGAGAAATGCTACCGCAGCCATGAAGAACCAGGTTGCAAGATTTAATGACCTCAGGTTTGTCGGTCGAAGTGGAAGAGGGAAAAGCTTCACTCTGACCATCACTGTCTTCACAAACCCACCGCAAGTCGCCACCTACCACAGAGCCATCAAAATCACAGTGGATGGGCCCCGAGAACCTCGAAGACATCGGCAGAAACTAGATGATCAGACCAAGCCCGGGAGCTTGTCCTTTTCCGAGCGGCTCAGTGAACTGGAGCAGCTGCGGCGCACAGCCATGAGGGTCAGCCCACACCACCCAGCCCCCACGCCCAACCCTCGTGCCTCCCTGAACCACTCCACTGCCTTTAACCCTCAGCCTCAGAGTCAGATGCAGGATACAAGGCAGATCCAACCATC
Ensembl | human accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
human ... RUNX1(861)
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| Ensembl | human accession no. | Ensembl | human accession no. | Ensembl | human accession no. | Ensembl | human accession no. |
| product line MISSION® | product line MISSION® | product line MISSION® | product line MISSION® |
| Gene Information human ... RUNX1(861) | Gene Information human ... RUNX2(860) | Gene Information human ... STC2(8614) | Gene Information human ... CCNB1(891) |
| esiRNA cDNA target sequence GGCTGGCAATGATGAAAACTACTCGGCTGAGCTGAGAAATGCTACCGCAGCCATGAAGAACCAGGTTGCAAGATTTAATGACCTCAGGTTTGTCGGTCGAAGTGGAAGAGGGAAAAGCTTCACTCTGACCATCACTGTCTTCACAAACCCACCGCAAGTCGCCACCTACCACAGAGCCATCAAAATCACAGTGGATGGGCCCCGAGAACCTCGAAGACATCGGCAGAAACTAGATGATCAGACCAAGCCCGGGAGCTTGTCCTTTTCCGAGCGGCTCAGTGAACTGGAGCAGCTGCGGCGCACAGCCATGAGGGTCAGCCCACACCACCCAGCCCCCACGCCCAACCCTCGTGCCTCCCTGAACCACTCCACTGCCTTTAACCCTCAGCCTCAGAGTCAGATGCAGGATACAAGGCAGATCCAACCATC | esiRNA cDNA target sequence GGTACCAGATGGGACTGTGGTTACTGTCATGGCGGGTAACGATGAAAATTATTCTGCTGAGCTCCGGAATGCCTCTGCTGTTATGAAAAACCAAGTAGCAAGGTTCAACGATCTGAGATTTGTGGGCCGGAGTGGACGAGGCAAGAGTTTCACCTTGACCATAACCGTCTTCACAAATCCTCCCCAAGTAGCTACCTATCACAGAGCAATTAAAGTTACAGTAGATGGACCTCGGGAACCCAGAAGGCACAGACAGAAGCTTGATGACTCTAAACCTAGTTTGTTCTCTGACCGCCTCAGTGATTTAGGGCGCATTCCTCATCCCAGTATGAGAGTAGGTGTCCCGCCTCAGAACCCACGGCCCTCCCTGAACTCTGCACCAAGTCCTTTTAATCCACAAGGACAGAGTCAGATTACAGACCCCAGGC | esiRNA cDNA target sequence CAGTTCATGACCCTGGCTTTGGTGTTGGCCACCTTTGACCCGGCGCGGGGGACCGACGCCACCAACCCACCCGAGGGTCCCCAAGACAGGAGCTCCCAGCAGAAAGGCCGCCTGTCCCTGCAGAATACAGCGGAGATCCAGCACTGTTTGGTCAACGCTGGCGATGTGGGGTGTGGCGTGTTTGAATGTTTCGAGAACAACTCTTGTGAGATTCGGGGCTTACATGGGATTTGCATGACTTTTCTGCACAACGCTGGAAAATTTGATGCCCAGGGCAAGTCATTCATCAAAGACGCCTTGAAATGTAAGGCCCACGCTCTGCGGCACAGGTTCGGCTGCATAAGCCGGAAGTGCCCGGCCATCAGGGAAATGGTGTCCCAGTTGCAGCGGGAATGCTACCTCAAGCACGACCTGTG | esiRNA cDNA target sequence TTGGTGTCACTGCCATGTTTATTGCAAGCAAATATGAAGAAATGTACCCTCCAGAAATTGGTGACTTTGCTTTTGTGACTGACAACACTTATACTAAGCACCAAATCAGACAGATGGAAATGAAGATTCTAAGAGCTTTAAACTTTGGTCTGGGTCGGCCTCTACCTTTGCACTTCCTTCGGAGAGCATCTAAGATTGGAGAGGTTGATGTCGAGCAACATACTTTGGCCAAATACCTGATGGAACTAACTATGTTGGACTATGACATGGTGCACTTTCCTCCTTCTCAAATTGCAGCAGGAGCTTTTTGCTTAGCACTGAAAATTCTGGATAATGGTGAATGGACACCAACTCTACAACATTACCTGTCATATACTGAAGAATCTCTTCTTCCAGTTATGCAGCACCTGGCTAAGAATGTAGTCATGGTAAATCAAGGACTTACAAAGCACATGACTGTCAAGAACAAGTATGCCACATCGAAGCATG |
| NCBI accession no. | NCBI accession no. | NCBI accession no. | NCBI accession no. |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| EHU124231-20UG | 04061831359565 |
| EHU124231-50UG | 04061828348145 |
