Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
20 μg | Usually ships in 3 weeks (4 to 6 weeks for custom orders). | € 212,00 |
50 μg | Usually ships in 3 weeks (4 to 6 weeks for custom orders). | € 382,00 |
About This Item
€ 212,00
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
AAGTCACAGGCCTGACTTCCAGACTAGGCTCTGCCCCTAAGGAAATCACCTTTCTGGTCTTAGTCTCCCCATCTGAAAATGGCATAACCAACCTACCTGCCTCAAATACAAGACCCTGCTTGTGAAGCGTCTCCTGGCTCAGACCAGTGCTAGAAGTGTCCAGTCTAGCAGCTCCTGGACCAAGACTCCGTGGCCATTCCCTTCCTCCTGCCTTCCATCCACCCCCTTACGATACAGAAGTTGAGATACAGAGAGGTGAAGGAACTTGTCCCAGGCCCTACAGCTAGTAAGTGATGGACCTGGAATTTAAGCCCAGGTAGACTGCCTCTAGAGACTGTGCTTACAACCCTTTTATCCTGTGCCTGTTCCC
GenBank accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
human ... TP53TG5(27296)
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| esiRNA cDNA target sequence AAGTCACAGGCCTGACTTCCAGACTAGGCTCTGCCCCTAAGGAAATCACCTTTCTGGTCTTAGTCTCCCCATCTGAAAATGGCATAACCAACCTACCTGCCTCAAATACAAGACCCTGCTTGTGAAGCGTCTCCTGGCTCAGACCAGTGCTAGAAGTGTCCAGTCTAGCAGCTCCTGGACCAAGACTCCGTGGCCATTCCCTTCCTCCTGCCTTCCATCCACCCCCTTACGATACAGAAGTTGAGATACAGAGAGGTGAAGGAACTTGTCCCAGGCCCTACAGCTAGTAAGTGATGGACCTGGAATTTAAGCCCAGGTAGACTGCCTCTAGAGACTGTGCTTACAACCCTTTTATCCTGTGCCTGTTCCC | esiRNA cDNA target sequence ACAGCAGGGTTTCCAAGATGCAAGATGAGAAACTGCGGGACGAGACAGAGCAGCCTGTGAGCAAAGTAATTGAGCGGAACCGTCTGAGAACGGTGTTAAAAAACTTGTCGCTCTTGAAGCTACTCAAGAGCTCAAACCGCCGGATCCAAGAACTGCATAAGCTGGCCAAAAGGTGTTGGCATTCACTGCTCAGTGTTCCAAAGATTCTCCGCATCTCCTCTGGGGAAAACAGTGCCTGCAATAAAACAAAACAAAATAATGAAGAGTTCCAGGAGATCGGGTGCTCCGAGAAGGAACTCAAGTCCAAGAAATTAGAGTCCACAGGGGACCCTAAGAAAAAAGAGTACAAGGAGTGGAAGTCCCAGGTGCAGTCAGGGATGAGGAACAAGGAGAAAACGTCATTGGCGGCAATGCCACGGAAAGAAAAGCATATAGAGCCTGAGGTTCCAAGGACATCGAGGGATGAT | esiRNA cDNA target sequence AAAAGGTGTCCCTTGGATCATAGTTGAAAATAATAGTTGCTGAGTAGCTATTCAGTACTAGTTGAGAATGACACATTTTTAAAGTAGCACCTTTGTGGGGCATACAAAATCATATTCACTTGTCTGTAATAAGCTTTCTTGACTTGCTCTGATAATGGAGATAATAGAACAATTTGGGTTTATGGTTTAAACCCTACAATTAGATAATTGGAGTTTCCTCCTGCCTTTCAGAAGA | esiRNA cDNA target sequence ACCAAAGCTGAGAAGCTCCAGCTGCTGAACCACCGGCCTGTGACTGCTGTGGAGATCCAGCTGATGGTGGAAGAGAGTGAAGAGCGGCTCACGGAGGAGCAGATTGAAGCTCTTCTCCACACCGTCACCAGCATTCTGCCTGCAGAGCCAGAGGCTGAGCAGAAGAAGAATACAAACAGCAATGTGGCAATGGACGAAGAGGACCCAGCATAGAAGAGCACAGCTGGCCCCGGCGTTTCATGAAGTCAGAAGGCCTGGCAGCCATTTCCTGGACGTTGAGAGGATTGTTTATTTGATTTTTATCCTCATCCCAGCAGGCCTGGCTTTGTGGTTAGTTGGGTACATCACAAAAATAAGTTAAAAAGAAATATTTGTGCCTTGGGGAGAAGAAACATGGTGAAAACAGGCTGAGG |
| product line MISSION® | product line MISSION® | product line MISSION® | product line MISSION® |
| Gene Information human ... TP53TG5(27296) | Gene Information human ... TP53TG5(27296), C20orf10(27296) | Gene Information mouse ... SOCS4(67296), Socs4(67296) | Gene Information human ... CRCP(27297), AC068533.6(27297) |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| shipped in ambient | shipped in ambient | shipped in ambient | shipped in ambient |
Storage Class
12 - Non Combustible Liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
wgk
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
