The presence of EDTA, SDS, or excess salt can affect my PCR/ sequencing reaction. The lysis buffer information is proprietary, but we can say it is free of chaotropic salts. The resins are desalting resins so EDTA, SDS, and salts are depleted.
Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 10 reactions | Check Cart for Availability | € 51,10 | |
| 50 reactions | Check Cart for Availability | € 241,00 | |
| 250 reactions | Check Cart for Availability | € 1.010,00 |
About This Item
purified by
(Single-spin negative chromotography), (Time: 45 minutes or less)
feature
Compatible Application (Suitable for most common downstream applications, including genotyping, PCR, and NGS), Intended use (For the purificiation of genomic DNA from human or animal tissue), Typical/expected yield (Varies by sample. Please reference user guide for more information.)
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
technique(s)
DNA purification: suitable
input
tissue (fresh or frozen)
test parameters
: 3 min hands on time, sample volume: 20 mg
greener alternative category
storage temp.
2-8°C
General description
- Isolation of DNA in under an hour with minimal sample handling
- Better purity, leading to improved performance in PCR and other applications
- Significant reduction in plastic and hazardous chemical waste
Traditional silica-based, bind-wash-elute purification kits require multiple wash steps to remove impurities from the spin columns. These steps increase the risk of cross contamination, subject the DNA to centrifugation sheering forces, and introduce chaotropic salts that can carry over into the final sample, inhibiting downstream applications.
GenElute™-E kits employ size exclusion negative chromatography to separate large nucleic acid molecules from smaller protein, lipid, and ionic components in cell, tissue, blood, and other samples. Single-spin columns efficiently absorb and retain cellular debris and sample contaminants while allowing nucleic acids to pass through, reducing the number of steps and plastic materials required for purification. This novel method for high-quality purification is made possible by our innovative SmartLyse® Protease, which enables fast and efficient lysis of a wide range of sample types.
Isolate nucleic acid in a fraction of time compared to traditional silica bind-wash-elute procedures. Simply mix sample with lysis components and incubate according to kit protocol, pipette sample onto the spin column, and centrifuge directly into a collection tube. Cellular debris and contaminants remain bound in the column to be discarded, while purified DNA is ready to be used in downstream applications or stored.
Application
The initial lysis step takes place under physiological conditions where the enzyme activity of proteases is generally increased. SmartLyse® Protease contains a unique combination of several active enzymes with an expanded substrate specificity. This eliminates the requirement for extended, overnight incubations, which are required in many conventional protocols.
Purified genomic DNA is eluted in Tris buffer, pH 7.8 and can immediately be used for most common downstream applications, including PCR, genotyping, NGS, and others. Final yield is equal or better than silica-based methods depending on sample type. GenElute™-E purified genomic DNA preparations commonly show an A260/280 ratio of around 1.8.
Features and Benefits
Preparation Note
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) DNA purification: suitable | technique(s) - | technique(s) DNA purification: suitable | technique(s) DNA purification: suitable |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| input tissue (fresh or frozen) | input - | input cells | input blood (liquid or dried) |
| feature Compatible Application (Suitable for most common downstream applications, including genotyping, PCR, and NGS), Typical/expected yield (Varies by sample. Please reference user guide for more information.), Intended use (For the purificiation of genomic DNA from human or animal tissue) | feature Compatible Application (Suitable for most common downstream applications, including genotyping, PCR, and NGS), Intended use (For the purificiation of genomic DNA from human or animal tissue), Typical/expected yield (Varies by sample. Please reference user guide for more information.) | feature Compatible Application (Suitable for most common downstream applications, including genotyping, PCR, and NGS), Intended use (For purification of genomic DNA from cell lines), Typical/expected yield (Varies by sample. Please reference user guide for more information.) | feature Compatible Application (Suitable for most common downstream applications, including genotyping, PCR, and NGS), Intended use (For the purification of genomic DNA from whole or dried blood samples), Typical/expected yield (Varies by sample. Please reference user guide for more information.) |
| test parameters : 3 min hands on time, sample volume: 20 mg | test parameters - | test parameters : 3 min hands on time | test parameters : 3 min hands on time, sample volume: 60 μL blood or 2-5 dried blood spots |
| sustainability Greener Alternative Product | sustainability Greener Alternative Product | sustainability Greener Alternative Product | sustainability Greener Alternative Product |
Kit Components Only
- Tissue Lysis Buffer
- SmartLyse® T Protease
- Clearing Solution T
- RNase A Tissue
- 1x Tris Buffer
- Spin Columns
signalword
Danger
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Auswirkungen der Aufreinigungsmethode auf die Genauigkeit der DNA-Quantifizierung und nachgeschaltete enzymatische Prozesse Bewertung der Reinheit genomischer DNA durch UV-Spektralphotometrie, Gelelektrophorese und nachgeschaltete qPCR bei Verwendung von GenElute™-E DNA-Aufreinigungskits.
GenElute™-E DNA purification kits ensure accurate DNA quantitation and downstream enzymatic processes.
Related Content
Demo video showing how to purify DNA and RNA GenElute™-E single spin nucleic acid purification kits
Answers to frequently asked questions related to GenElute™-E single spin DNA and RNA purification and negative chromatography
Reduce plastic waste and eliminate hazardous liquid waste for more sustainable laboratories with GenElute™-E Single Spin DNA and RNA prep kits.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| EC300-10RXN | 04061842202683 |
| EC300-50RXN | 04061842202690 |
| EC300-250RXN | 04061842202706 |
-
What is the composition of the lysis buffer and clearing buffer after flowing through the resin?
1 answer-
Helpful?
-
-
Genelute-e single spin DNA and RNA purification kits use negative chromatographty to isolate nucleic acids. Can you explain how this approach simplifies workflows?
1 answer-
Instead of optimizing the bind, wash, and release steps in conventional silica-based spin purification preps, the technology focuses on a separation by performing a single step fractionation based on the size of the biomolecules, which results in depleted impurities.
Helpful?
-
-
Does the technology introduce any bias into the sample?
1 answer-
GenElute™-E does not introduce biases that some “bind-wash-elute” technologies can add because the technology separates by size, rather than by what binds and what is released.
Helpful?
-
-
Do we know how stable the purified DNA is through several freeze-thaw cycles?
1 answer-
This will fluctuate due to sample variability (sample collection, concentration, fragment length, sequence [GC content], storage before isolation, etc.). However, the sample is buffer exchanged into a standard storage buffer that is included in the kit (1X TE).
Helpful?
-





