Skip to Content
Merck
All Photos(1)

Key Documents

E8755

Sigma-Aldrich

Erythromycin ethyl succinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H75NO16
CAS Number:
Molecular Weight:
862.05
EC Number:
MDL number:
UNSPSC Code:
51282328
PubChem Substance ID:
NACRES:
NA.85

biological source

microbial

form

solid

potency

≥780 μg per mg

color

white

solubility

ethanol: 50 mg/mL

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

protein synthesis | interferes

SMILES string

CCOC(=O)CCC(=O)OC1C(CC(C)OC1OC2C(C)C(OC3CC(C)(OC)C(O)C(C)O3)C(C)C(=O)OC(CC)C(C)(O)C(O)C(C)C(=O)C(C)CC2(C)O)N(C)C

InChI

1S/C43H75NO16/c1-15-29-43(11,52)36(48)24(5)33(47)22(3)20-41(9,51)38(25(6)34(26(7)39(50)57-29)59-32-21-42(10,53-14)37(49)27(8)56-32)60-40-35(28(44(12)13)19-23(4)55-40)58-31(46)18-17-30(45)54-16-2/h22-29,32,34-38,40,48-49,51-52H,15-21H2,1-14H3

InChI key

NSYZCCDSJNWWJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide

Application

Erythromycin ethyl succinate is used to study erythromycin-resistant Streptococcus pyogenes and Streptococcus pneumoniae[1]. It has been used to study down-regulation of motilin receptors on rabbit colon myocytes[2] and lethal mutations in outer membrane genes omsA and firA in Salmonella typhimurium[3].

Biochem/physiol Actions

Erythromycin inhibits protein synthesis (elongation) at the level of transpeptidation (aminoacyl translocation A-site to P-site) by binding to the 50s subunit of the bacterial 70s rRNA complex.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Vuorio et al.
Journal of bacteriology, 174(22), 7090-7097 (1992-11-01)
We have previously identified the gene (the ssc gene) defective in the thermosensitive and antibiotic-supersusceptible outer membrane permeability mutant SS-C of Salmonella typhimurium and shown that this gene is analogous to the Escherichia coli gene firA (L. Hirvas, P. Koski
S D Bologna et al.
The Journal of pharmacology and experimental therapeutics, 266(2), 852-856 (1993-08-01)
Acutely, erythromycin stimulates colonic smooth muscle contraction via action on motilin receptors, but the effects of chronic erythromycin exposure are unknown. Thus contraction and motilin binding studies were performed on rabbit colonic smooth muscle after 2 weeks of oral erythromycin
K P Klugman et al.
The Journal of antimicrobial chemotherapy, 42(6), 729-734 (1999-03-03)
The susceptibility of 40 erythromycin-resistant isolates of Streptococcus pyogenes and 40 multiply-resistant isolates of Streptococcus pneumoniae to six macrolide antibiotics, representing 14-, 15- and 16-membered lactone ring structures, was tested. The genetic basis for macrolide resistance in the strains was
J J Stewart et al.
Journal of clinical pharmacology, 40(4), 347-353 (2000-04-13)
Motion sickness and the antimotion sickness drugs scopolamine (SCP) and promethazine (PMZ) inhibit gastric emptying (GE). This study was conducted to determine if erythromycin would exert its well-known prokinetic effects in normal and motion-sick subjects given antimotion sickness drugs. Fifteen
Y A Cho et al.
Die Pharmazie, 67(2), 124-130 (2012-04-20)
The effects of curcumin, a natural anti-cancer compound, on the bioavailability and pharmacokinetics of tamoxifen and its metabolite, 4-hydroxytamoxifen, were investigated in rats. Tamoxifen and curcumin interact with cytochrom P450 (CYP) enzymes and P-glycoprotein, and the increase in the use

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service