Skip to Content
Merck

Skip To

E3539

Proteinase 3 from human leukocytes

≥95% (PAGE)

Synonym(s):

AGP7, Myeloblastin, P29B, PR-3, PR3 Ag

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
MDL number:
Assay:
≥95% (PAGE)


assay

≥95% (PAGE)

form

buffered aqueous solution

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... PRTN3(5657)

General description

Proteinase 3 is found in high concentration in the primary granules of myeloid leukemia blasts.4

Application

Proteinase 3 has been shown to be important in the pathogenesis of human emphysema.1 It has also been used in a study to investigate the liberation of the antibacterial and cytotoxic peptide LL-37 by cleavage of human cathelicidin hCAP-183.2

Biochem/physiol Actions

Proteinase 3 (PR-3) is a serine protease expressed in the granules of neutrophils and monocytes. Also known as myeloblastin and p29b, it degrades elastin and is associated with myeloid leukemia and Wegener′s granulomatosis.3
Used for the detection of anti-PR3 auto-antibodies in immunoassays.

Physical form

Solution in 50 mM sodium acetate (pH 5) with 600 mM sodium chloride


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
E3539-100UG04061832700687

Questions

Reviews

No rating value

Active Filters