Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C32H37N5O5
CAS Number:
Molecular Weight:
571.67
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
storage temp.
−20°C
SMILES string
N[C@@H](Cc1ccc(O)cc1)C(=O)N2CCC[C@H]2C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](Cc4ccccc4)C(N)=O
InChI
1S/C32H37N5O5/c33-25(18-23-13-15-24(38)16-14-23)32(42)37-17-7-12-28(37)31(41)36-27(20-22-10-5-2-6-11-22)30(40)35-26(29(34)39)19-21-8-3-1-4-9-21/h1-6,8-11,13-16,25-28,38H,7,12,17-20,33H2,(H2,34,39)(H,35,40)(H,36,41)/t25-,26-,27-,28-/m0/s1
InChI key
XIJHWXXXIMEHKW-LJWNLINESA-N
Gene Information
human ... OPRM1(4988)
mouse ... OPRM1(18390)
rat ... OPRM1(25601)
General description
Endomorphin 2 is an endogenous ligand for μ-opioid receptor (MOR). It is a neuropeptide and has the sequence Tyr-Pro-Phe-Phe-NH2. It has a greater affinity for MOR1 receptor than MOR2.[1] It resides in primary sensory afferent fibers, and might be the predominant ligand to regulate pain perception.[2] In dynorphin-induced allodynia mice models, endomorphin has anti-allodynic effects. In tail flick test, it produces short-lived antinociception, which is naloxone-sensitive.[3]
Biochem/physiol Actions
Potent, selective endogenous μ opioid receptor agonist.
Legal Information
Sold under license to US patent number 6,303,578
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
I E Goldberg et al.
The Journal of pharmacology and experimental therapeutics, 286(2), 1007-1013 (1998-08-08)
The recently isolated peptides endomorphin-1 and endomorphin-2 have been suggested to be the endogenous ligands for the mu receptor. In traditional opioid receptor binding assays in mouse brain homogenates, both endomorphin-1 and endomorphin-2 competed both mu1 and mu2 receptor sites
Gianfranco Balboni et al.
Journal of medicinal chemistry, 49(18), 5610-5617 (2006-09-01)
Substitution of Gly with side-chain-protected or unprotected Lys in lead compounds containing the opioid pharmacophore Dmt-Tic [H-Dmt-Tic-Gly-NH-CH(2)-Ph, mu agonist/delta antagonist; H-Dmt-Tic-Gly-NH-Ph, mu agonist/delta agonist; and H-Dmt-Tic-NH-CH(2)-Bid, delta agonist (Bid = 1H-benzimidazole-2-yl)] yielded a new series of compounds endowed with distinct
Masami Tanaka et al.
PloS one, 4(5), e5532-e5532 (2009-05-15)
MicroRNAs are a family of 19- to 25-nucleotides noncoding small RNAs that primarily function as gene regulators. Aberrant microRNA expression has been described for several human malignancies, and this new class of small regulatory RNAs has both oncogenic and tumor