Skip to Content
Merck

Skip To

E2153

E-Cadherin/Fc Chimera from mouse

>90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym(s):

L-CAM, Arc-1, Cell-CAM120/80, Epithelial cadherin, Uvomorulin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
50 μg
Check Cart for Availability
€ 787,00
€ 668,95

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
MDL number:
Form:
lyophilized powder
Assay:
>90% (SDS-PAGE)
Biological source:
mouse
Recombinant:
expressed in NSO cells
Mol wt:
monomer calculated mol wt 88.2 kDa, ~120 kDa by SDS-PAGE (reducing), calculated mol wt 88.2 kDa

€ 668,95

List Price€ 787,00Save 15%
Web-Only Promotion

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

recombinant

expressed in NSO cells

sterility

sterile; sterile-filtered

assay

>90% (SDS-PAGE)

form

lyophilized powder

mol wt

monomer calculated mol wt 88.2 kDa, ~120 kDa by SDS-PAGE (reducing), calculated mol wt 88.2 kDa

packaging

pkg of 50 μg

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

mouse ... Cdh1(12550)

General description

E (epithelial)-cadherin is a type I classical cadherin, which exists as a dimer. It has an ectodomain made of five extracellular cadherin-like (EC) domains, connected by conserved linker regions, which are calcium-binding in nature. It spans the membrane once, and contains a cytoplasmic region.[1] It is a glycoprotein, and in humans has a molecular weight of 120kDa. It binds to catenin and cytoskeletal protein with its C-terminal domain.[2]
Extracellular domain of mouse E-Cadherin (amino acids 1-709) fused to the carboxy-terminal 6X histidine tagged Fc region of human IgG1.

Application

E-Cadherin/Fc Chimera from mouse has been used in adhesion assays.

Biochem/physiol Actions

E (epithelial)-cadherin is involved in hemophilic cell-cell adhesion in a calcium-dependent manner. It forms adheren junctions through its ectodomains, to maintain physical contacts between adjacent cells.[1] It plays essential roles in developmental processes, tissue homeostasis and multiple diseases. In humans, this protein might be involved in the pathophysiology of ulcerative colitis. In Crohn′s diseased patients, mutations in this gene lead to a truncated or mislocalized protein product.[3] Epithelial-mesenchymal transition (EMT) and mesenchymal-epithelial transition (MET) are the underlying processes of tumorigenesis and embryogenesis, and these involve the control of E-cadherin expression.[4]

Physical form

Lyophilized from 0.2 μm filtered solution in 20mM MES, 300mM NaCl, 1mM CaCl2.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E0778E0528N6784
biological source

mouse

biological source

mouse

biological source

human

biological source

mouse

assay

>90% (SDS-PAGE)

assay

>90% (SDS-PAGE)

assay

>90% (SDS-PAGE)

assay

>95% (SDS-PAGE)

recombinant

expressed in NSO cells

recombinant

expressed in NSO cells

recombinant

expressed in NSO cells

recombinant

expressed in NSO cells

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

-

technique(s)

-

mol wt

monomer calculated mol wt 88.2 kDa, ~120 kDa by SDS-PAGE (reducing)

mol wt

monomer calculated mol wt 49.6 kDa, 60-65 kDa by SDS-PAGE (reducing)

mol wt

monomer calculated mol wt 48.6 kDa, 50-55 kDa by SDS-PAGE (reducing)

mol wt

55-60 kDa by SDS-PAGE (reducing), calculated mol wt 50 kDa

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Instructions


Xenopus δ-catenin is essential in early embryogenesis and is functionally linked to cadherins and small GTPases.
Gu D, et al.
Journal of Cell Science, 122(22), 4049-4061 (2009)
Dongmin Gu et al.
Journal of cell science, 122(Pt 22), 4049-4061 (2009-10-22)
Catenins of the p120 subclass display an array of intracellular localizations and functions. Although the genetic knockout of mouse delta-catenin results in mild cognitive dysfunction, we found severe effects of its depletion in Xenopus. delta-catenin in Xenopus is transcribed as
Ying Li et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(41), 16462-16467 (2013-09-27)
Epithelial cadherin (E-cadherin), a member of the classical cadherin family, mediates calcium-dependent homophilic cell-cell adhesion. Crystal structures of classical cadherins reveal an adhesive dimer interface featuring reciprocal exchange of N-terminal β-strands between two protomers. Previous work has identified a putative



Global Trade Item Number

SKUGTIN
E2153-50UG04061833602478

Questions

Reviews

No rating value

Active Filters