Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
12352202
NACRES:
NA.77
Form:
lyophilized powder
Mol wt:
7.3 kDa
Pricing and availability is not currently available.
Quality Level
form
lyophilized powder
mol wt
7.3 kDa
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
-10 to -25°C
General description
Recombinant human Epidermal Growth Factor is small polypeptide of 6.2 kDa protein containing 53 amino acid residues. The EGF gene encodes for a large type I membrane–anchored precursor protein which is cleaved into pro-EGF for generation of peptide hormone EGF. EGF can be found in human platelets, macrophages, urine, saliva, milk, and plasma.
Biochem/physiol Actions
Epidermal Growth Factor (EGF) is a growth factor that plays an important role in the regulation of cell growth, proliferation, and differentiation by binding to its receptor EGFR. Biological activities ascribed to EGF include epithelial development, angiogenesis, inhibition of gastric acid secretion, fibroblast proliferation, and colony formation of epidermal cells in culture.
Preparation Note
For reconstitution: Briefly centrifuge the vial before opening. Reconstitute in water to a concentration of 0.1 mg/mL. Do not vortex. This solution is recommended to store in working aliquots at –20 oC to -80 oC. Store the lyophilized product at –20 oC. The product is stable for at least 2 years as supplied.
Still not finding the right product?
Explore all of our products under EGF human
Storage Class
11 - Combustible Solids
wgk
WGK 2
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Discover validated bioactivity and proliferation data for our portfolio of animal component-free cell culture growth factors.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| E1235-10UG | 04065271256620 |
| E1235-50UG | 04065271256637 |