Skip to Content
Merck
All Photos(1)

Key Documents

E0643

Sigma-Aldrich

Erythropoietin Soluble Receptor Peptide

>90% (SDS-PAGE), lyophilized powder, human recombinant, expressed in NSO cells

Synonym(s):

EPO sR

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€936.00

€936.00


Estimated to ship on25 May 2025



Select a Size

Change View
50 μG
€936.00

About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

€936.00


Estimated to ship on25 May 2025


Product Name

Erythropoietin Soluble Receptor human, >90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

biological source

human

Quality Level

recombinant

expressed in NSO cells

Assay

>90% (SDS-PAGE)

form

lyophilized powder

potency

74 ng/mL

mol wt

~25 kDa

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... EPOR(2057)

Biochem/physiol Actions

Binds erthropoietin with high affinity and is an erythropoietin antagonist.
Recombinant human soluble erythropoietin receptor binds erthropoietin with high affinity and is an erythropoietin antagonist.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 5% trehalose and 50 ug of bovine serum albumin per 1 ug of cytokine.

Analysis Note

The biological activity is measured by its ability to inhibit the recombinant human erythropoietin mediated cell proliferation of human TF1 cells.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soluble Erythropoietin Receptor Contributes To Erythropoietin Resistance In End-stage Renal Disease.
Khankin, E.V., et al.
PLoS Biology, 5, e9246-e9246 (2010)
T Kitamura et al.
Journal of cellular physiology, 140(2), 323-334 (1989-08-01)
We have established a novel cell line, designated as TF-1, from a patient with erythroleukemia, which showed complete growth dependency on granulocyte-macrophage colony-stimulating factor (GM-CSF) or on interleukin-3 (IL-3) and carried a homogeneous chromosomal abnormality (54X). Erythropoietin (EPO) also sustained

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service