Skip to Content
Merck
All Photos(1)

Key Documents

658552

Sigma-Aldrich

AG 1478

A cell-permeable, reversible, ATP-competitive, highly potent and selective inhibitor of epidermal growth factor receptor kinase versus HER2-neu and platelet-derived growth factor receptor kinase.

Synonym(s):

AG 1478, 4-(3-Chloroanilino)-6,7-dimethoxyquinazoline

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€235.00

€235.00


Estimated to ship on09 April 2025



Select a Size

Change View
5 MG
€235.00

About This Item

Empirical Formula (Hill Notation):
C16H14ClN3O2
CAS Number:
Molecular Weight:
315.75
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€235.00


Estimated to ship on09 April 2025


Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Clc1cc(ccc1)Nc2ncnc3c2cc(c(c3)OC)OC

InChI

1S/C16H14ClN3O2/c1-21-14-7-12-13(8-15(14)22-2)18-9-19-16(12)20-11-5-3-4-10(17)6-11/h3-9H,1-2H3,(H,18,19,20)

InChI key

GFNNBHLJANVSQV-UHFFFAOYSA-N

General description

A cell-permeable, reversible, ATP-competitive, highly potent and selective inhibitor of epidermal growth factor receptor kinase (IC50 = 3 nM) versus HER2-neu (IC50 >100 µM) and platelet-derived growth factor receptor kinase (IC50 >100 µM). Abolishes MAP kinase (ERK) activation induced by Angiotensin II (Cat. No. 05-23-0101). Also inhibits the activation of EGFR kinase and MAP kinase by 4-hydroxynonenal. Downregulates ARF1 activity and disperses Golgi structure. A 10 mM (1 mg/317 µl) solution of AG 1478 (Cat. No. 658548) in DMSO is also available.
A cell-permeable, reversible, ATP-competitive, highly potent and specific inhibitor of the epidermal growth factor (EGF) receptor tyrosine kinase (IC50 = 3 nM). Much higher concentrations are required for inhibition of the kinase activity of the closely-related HER2 (neu/erb-B2) receptor (IC50 >100 µM), the platelet-derived growth factor (PDGF) receptor (IC50 >100 µM), and p210Bcr-Abl (IC50 >50 µM). Its mechanism of inhibition likely mimics that of the related compound, 4-(3-chloroanilino)quinazoline (CAQ), a competitive inhibitor with respect to ATP. Downregulates ARF1 activity and disperses Golgi structure.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Epidermal growth factor receptor kinase
Product competes with ATP.
Reversible: yes
Target IC50: 3 nM against epidermal growth factor receptor kinase

Warning

Toxicity: Standard Handling (A)

Preparation Note

Further dilute with aqueous buffers just prior to use.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Pan, H., et al. 2008. J. Biol. Chem.283, In press.
Liu, W., et al. 1999. J. Cell Sci.112, 2409.
Eguchi, S., et al. 1998. J. Biol. Chem. 273, 8890.
Levitzki, A., and Gazit, A. 1995. Science267, 1782.
Fry, D.W., et al. 1994. Science265, 1093.
Osherov, N., and Levitski, A. 1994. Eur. J. Biochem.225, 1047.
Ward, W.H., et al. 1994. Biochem. Pharmacol.48, 659.

Legal Information

Sold under license of U.S. Patent 5,457,105 and European Patent 0,566,266.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandro Brombin et al.
Cell reports, 38(2), 110234-110234 (2022-01-13)
Melanocytes, the pigment-producing cells, are replenished from multiple stem cell niches in adult tissue. Although pigmentation traits are known risk factors for melanoma, we know little about melanocyte stem cell (McSC) populations other than hair follicle McSCs and lack key
S P Soltoff
The Journal of biological chemistry, 273(36), 23110-23117 (1998-08-29)
The activation of growth factor receptors and receptors coupled to heterotrimeric guanine nucleotide-binding proteins (G-proteins) can increase mitogen-activated protein (MAP) kinase activity in many cells. Previously, we demonstrated that the activation of G-protein-coupled P2Y2 receptors by extracellular ATP and UTP
Tong San Tan et al.
Journal of cell science, 134(19) (2021-10-14)
In the skin fragility disorder epidermolysis bullosa simplex (EBS), mutations in keratin 14 (K14, also known as KRT14) or keratin 5 (K5, also known as KRT5) lead to keratinocyte rupture and skin blistering. Severe forms of EBS are associated with
Akito Kakiuchi et al.
Oncology reports, 45(4) (2021-03-03)
In human head and neck squamous cell carcinoma (HNSCC), the invasion and metastatic properties of cancer cells are promoted by junctional adhesion molecule‑A (JAM‑A) and claudin‑1; these are epithelial tight junction molecules regulated by histone deacetylases (HDACs) and transcription factor
Zachary M Harris et al.
Oxidative medicine and cellular longevity, 2022, 9518592-9518592 (2022-10-05)
Studies have linked severe hyperoxia, or prolonged exposure to very high oxygen levels, with worse clinical outcomes. This study investigated the role of epidermal growth factor receptor (EGFR) in hyperoxia-induced lung injury at very high oxygen levels (>95%). Effects of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service