Skip to Content
Merck

Skip To

E0159

Efonidipine hydrochloride monoethanolate

≥98% (HPLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 96,40
€ 81,94

About This Item

Empirical Formula (Hill Notation):
C34H38N3O7P · HCl · C2H5OH
CAS Number:
Molecular Weight:
714.18
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated

€ 81,94

List Price€ 96,40Save 15%
Web-Only Promotion

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

pale yellow

solubility

DMSO: >5 mg/mL, H2O: insoluble

originator

Shionogi

storage temp.

2-8°C

SMILES string

Cl.CCO.CC1=C(C(c2cccc(c2)[N+]([O-])=O)C(=C(C)N1)P3(=O)OCC(C)(C)CO3)C(=O)OCCN(Cc4ccccc4)c5ccccc5

InChI

1S/C34H38N3O7P.C2H6O.ClH/c1-24-30(33(38)42-19-18-36(28-15-9-6-10-16-28)21-26-12-7-5-8-13-26)31(27-14-11-17-29(20-27)37(39)40)32(25(2)35-24)45(41)43-22-34(3,4)23-44-45;1-2-3;/h5-17,20,31,35H,18-19,21-23H2,1-4H3;3H,2H2,1H3;1H

InChI key

IKBJGZQVVVXCEQ-UHFFFAOYSA-N

Biochem/physiol Actions

Efonidipine hydrochloride is a racemic compound that has some L-type as well as T-type calcium channel blocking activity.
Racemic efonidipine has both L-type and T-type calcium channel blocking activity. The R(−)-isomer appears to be very selective for T-type calcium channel. The S(+)-isomer inhibits the expressed Ca(V)1.2, Ca(V)1.3 and Ca(V)3.1 channel currents almost equally.

Features and Benefits

This compound was developed by Shionogi. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML0346SML0807SML1261
form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: >5 mg/mL, H2O: insoluble

solubility

H2O: ≥5 mg/mL at warmed

solubility

DMSO: 5 mg/mL, clear (warmed)

solubility

DMSO: 5 mg/mL, clear (warmed)

storage condition

desiccated

storage condition

desiccated

storage condition

desiccated

storage condition

desiccated


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters