About This Item
Quality Segment
assay
≥98% (HPLC)
form
solid
color
white to off-white
solubility
DMSO: >10 mg/mL
SMILES string
CCN1C(=O)Nc2cc(Cl)c(Cl)cc12
InChI
1S/C9H8Cl2N2O/c1-2-13-8-4-6(11)5(10)3-7(8)12-9(13)14/h3-4H,2H2,1H3,(H,12,14)
InChI key
LKHRMULASXZCLG-UHFFFAOYSA-N
Gene Information
Biochem/physiol Actions
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥98% (HPLC) | assay ≥98% (HPLC) | assay - | assay ≥98% (HPLC) |
| form solid | form solid | form solid | form solid |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 200 |
| solubility DMSO: >10 mg/mL | solubility DMSO: >10 mg/mL | solubility DMSO: soluble 28 mg/mL | solubility H2O: ≥2 mg/mL, DMSO: >20 mg/mL |
| color white to off-white | color white | color light yellow | color white |
| Gene Information human ... HTR1A(3350), HTR1B(3351), HTR1D(3352), HTR1E(3354), HTR1F(3355), HTR2A(3356), HTR2B(3357), HTR2C(3358), HTR3A(3359), HTR3B(9177), HTR3C(170572), HTR3D(200909), HTR3E(285242), HTR4(3360), HTR5A(3361), HTR5B(645694), HTR6(3362), HTR7(3363) | Gene Information human ... ADRB1(153), ADRB2(154), ADRB3(155) | Gene Information human ... PDE3A(5139), PDE3B(5140) | Gene Information - |
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




