Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
50 ea | Check Cart for Availability | € 1.170,00 € 1.053,00 |
100 ea | Check Cart for Availability | € 1.740,00 |
About This Item
NACRES:
NB.22
UNSPSC Code:
41123100
€ 1.053,00
List Price€ 1.170,00Save 10%Web-Only Promotion
Check Cart for Availability
packaging
case of 100 ea, pack of 50 ea
technique(s)
analytical sample preparation: suitable
pore size
10 kDa MWCO
application(s)
life science and biopharma
General description
A unique disposable unit for small sample dialysis, 5-100 μL. Dialysis is rapid with the ultra thin membrane of regenerated cellulose. After dialysis, sample can be recovered, almost 100% by centrifugation in a microcentrifuge tube. Recommended for salt removal, buffer exchange, protein binding assays and as a equilibrium dialyzer.
The Micro DispoDIALYZER includes a membrane, foam float, cap and two collection tubes (1.5 mL).
The Micro DispoDIALYZER includes a membrane, foam float, cap and two collection tubes (1.5 mL).
1 of 1
This Item | |||
|---|---|---|---|
| description MWCO 10 kDa | description MWCO 1 kDa | description MWCO 2 kDa | description MWCO 50 kDa |
| technique(s) analytical sample preparation: suitable | technique(s) analytical sample preparation: suitable | technique(s) - | technique(s) - |
| application(s) life science and biopharma | application(s) life science and biopharma | application(s) - | application(s) - |
| packaging case of 100 ea, pack of 50 ea | packaging pack of 50 ea | packaging pack of 50 ea, case of 100 ea | packaging - |
| pore size 10 kDa MWCO | pore size 1 kDa MWCO | pore size 2 kDa MWCO | pore size 50 kDa MWCO |
Still not finding the right product?
Explore all of our products under Dispo-Biodialyzer
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
