Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
50 ea | Check Cart for Availability | € 1.200,00 € 1.140,00 |
100 ea | Check Cart for Availability | € 1.780,00 |
About This Item
NACRES:
NB.22
UNSPSC Code:
41123100
€ 1.140,00
List Price€ 1.200,00Save 5%Web-Only Promotion
Check Cart for Availability
packaging
case of 100 ea, pack of 50 ea
pore size
2 kDa MWCO
General description
A unique disposable unit for small sample dialysis, 5-100 μL. Dialysis is rapid with the ultra thin membrane of regenerated cellulose. After dialysis, sample can be recovered, almost 100% by centrifugation in a microcentrifuge tube. Recommended for salt removal, buffer exchange, protein binding assays and as a equilibrium dialyzer.
The Micro DispoDIALYZER includes a membrane, foam float, cap and two collection tubes (1.5 mL).
The Micro DispoDIALYZER includes a membrane, foam float, cap and two collection tubes (1.5 mL).
1 of 1
This Item | |||
|---|---|---|---|
| packaging case of 100 ea, pack of 50 ea | packaging pack of 50 ea | packaging case of 100 ea, pack of 50 ea | packaging - |
| pore size 2 kDa MWCO | pore size 1 kDa MWCO | pore size 10 kDa MWCO | pore size 50 kDa MWCO |
Still not finding the right product?
Explore all of our products under Dispo-Biodialyzer
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
