Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
50 ea | Check Cart for Availability | € 1.130,00 |
About This Item
NACRES:
NB.22
UNSPSC Code:
41123100
€ 1.130,00
Check Cart for Availability
pore size
50 kDa MWCO
General description
A unique disposable unit for small sample dialysis, 5-100 μL. Dialysis is rapid with the ultra thin membrane of regenerated cellulose. After dialysis, sample can be recovered, almost 100% by centrifugation in a microcentrifuge tube. Recommended for salt removal, buffer exchange, protein binding assays and as a equilibrium dialyzer.
The Micro DispoDIALYZER includes a membrane, foam float, cap and two collection tubes (1.5 mL).
The Micro DispoDIALYZER includes a membrane, foam float, cap and two collection tubes (1.5 mL).
1 of 1
This Item | |||
|---|---|---|---|
| pore size 50 kDa MWCO | pore size 1 kDa MWCO | pore size 2 kDa MWCO | pore size 1 kDa MWCO |
Still not finding the right product?
Explore all of our products under Dispo-Biodialyzer
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

