Skip to Content
Merck

Skip To

D8571

DNSP-11 Peptide

≥98% (HPLC), powder

Synonym(s):

Dopamine neuron stimulating peptide-11

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C50H81N15O18 · xC2HF3O2
Molecular Weight:
1180.27 (free base basis)
UNSPSC Code:
51111800
NACRES:
NA.32
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

DNSP-11 trifluoroacetate salt, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

mol wt

1180.27 Da

packaging

pkg of 1 mg

color

white to off-white

solubility

H2O: >2 mg/mL

storage temp.

−20°C

Biochem/physiol Actions

DNSP-11 is an 11-mer peptide from the pro-GDNF domain with neurotrophic and neuroprotective activity toward dopamine neurons.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
V7754P1614K2644
assay

≥98% (HPLC)

assay

≥97% (HPLC)

assay

≥90% (HPLC)

assay

≥95% (HPLC)

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

100

form

powder

form

lyophilized powder

form

powder

form

film

packaging

pkg of 1 mg

packaging

-

packaging

-

packaging

-

mol wt

1180.27 Da

mol wt

calculated mol wt 1524.78 Da

mol wt

calculated mol wt 1928

mol wt

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
D8571-1MG04061833589748

Questions

Reviews

No rating value

Active Filters