Skip to Content
Merck

Skip To

D6884

1,2:4,5-Di-O-isopropylidene-β-D-fructopyranose

Synonym(s):

Diacetone-D-fructose

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352200


storage temp.

−20°C





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library