Skip to Content
Merck

Skip To

D6760

Dimethyl yellow

Synonym(s):

4-(Dimethylamino)azobenzene, N,N-Dimethyl-4-(phenylazo)aniline, Butter yellow, Methyl yellow

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
C6H5N=NC6H4N(CH3)2
CAS Number:
Molecular Weight:
225.29
UNSPSC Code:
12161500
EC Number:
200-455-7
Beilstein/REAXYS Number:
746016
Colour Index Number:
11020
MDL number:


grade

indicator grade

technique(s)

titration: suitable

mp

111 °C (dec.) (lit.)

SMILES string

CN(C)c1ccc(cc1)N=Nc2ccccc2

InChI

1S/C14H15N3/c1-17(2)14-10-8-13(9-11-14)16-15-12-6-4-3-5-7-12/h3-11H,1-2H3

InChI key

JCYPECIVGRXBMO-UHFFFAOYSA-N



pictograms

Skull and crossbonesHealth hazard

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

60-11-7

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library