Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
EC Number:
296-281-4
NACRES:
NA.25
Pricing and availability is not currently available.
biological source
whole wheat flour
Quality Segment
assay
≥93% (TLC)
form
lyophilized powder
functional group
ester
shipped in
ambient
storage temp.
−20°C
SMILES string
O1C(C(C(C(C1CO)O)O)O)OCCCOCCCOC2OC(C(C(C2O)O)O)CO
InChI
1S/C18H34O13/c19-7-9-11(21)13(23)15(25)17(30-9)28-5-1-3-27-4-2-6-29-18-16(26)14(24)12(22)10(8-20)31-18/h9-26H,1-8H2
InChI key
FROLUYNBHPUZQU-UHFFFAOYSA-N
1 of 1
This Item | |||
|---|---|---|---|
| functional group ester | functional group ester | functional group - | functional group - |
| biological source whole wheat flour | biological source synthetic (organic) | biological source synthetic (organic) | biological source natural (organic) |
| assay ≥93% (TLC) | assay ≥99% (TLC) | assay ≥93% (TLC) | assay ≥93% (HPLC) |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| form lyophilized powder | form powder | form powder | form powder |
| shipped in ambient | shipped in ambient | shipped in dry ice | shipped in ambient |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| D4651-10MG | 04061833566541 |
| D4651-1MG | 04061833573761 |



