Skip to Content
Merck

Skip To

D3943

DAPH

≥98% (HPLC), solid

Synonym(s):

4,5-Dianilinophthalimide, 5,6-Bis(phenylamino)-1H-isoindole-1,3(2H)-dione, CGP 52411

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Check Cart for Availability
€ 607,00

About This Item

Empirical Formula (Hill Notation):
C20H15N3O2
CAS Number:
Molecular Weight:
329.35
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
51111800
MDL number:
Assay:
≥98% (HPLC)
Form:
solid

€ 607,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

solid

color

orange

solubility

DMSO: >20 mg/mL, H2O: insoluble

storage temp.

2-8°C

SMILES string

O=C1NC(=O)c2cc(Nc3ccccc3)c(Nc4ccccc4)cc12

InChI

1S/C20H15N3O2/c24-19-15-11-17(21-13-7-3-1-4-8-13)18(12-16(15)20(25)23-19)22-14-9-5-2-6-10-14/h1-12,21-22H,(H,23,24,25)

InChI key

AAALVYBICLMAMA-UHFFFAOYSA-N

Gene Information

human ... EGFR(1956)

Biochem/physiol Actions

DAPH is an inhibitor of Aβ fibril formation; Protein tyrosine kinase inhibitor, specific for EGFR.

Features and Benefits

This compound is featured on the EGFR page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
H9415SML2418SML1013
form

solid

form

solid

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: >20 mg/mL, H2O: insoluble

solubility

DMSO: >20 mg/mL

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

color

orange

color

white to off-white

color

white to beige

color

white to gray


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters